Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00477486_m1
          See other AMBN GE Assays ›
          Gene Symbol:
          Ambn
          Gene Name:
          ameloblastin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 88455991 - 88468531 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00477486_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ambn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK017250.1 - 3 - 4 251 57
          BC087927.1 - 3 - 4 251 30504437 57
          U65021.1 - 3 - 4 231 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000811.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999463 U65021.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999463 AK017250.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999463 NM_009664.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 506785 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 503917 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 198847 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 507933 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 233270 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 407626 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 765597 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 37662 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151637 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P135359 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999463 U65021.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999463 AK017250.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999463 NM_009664.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P340484 U65021.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P340484 AK017250.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P340484 NM_009664.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P340484 BC087927.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872250 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872268 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872255 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872265 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872271 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872264 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872266 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872247 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872253 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P135359 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01155063 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151637 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05646747 - -
          Whole Mouse Genome G4122A A_51_P340484 U65021.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P340484 AK017250.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P340484 NM_009664.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P340484 BC087927.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113122 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139818 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151637 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P135359 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04436308 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04436305 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04436306 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04436319 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04436313 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04436332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04694832 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872390 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872378 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04429470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04436333 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872255 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872376 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872244 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04436317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872391 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872281 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04814144 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04219249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872279 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04122571 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872243 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872248 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872283 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872271 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04436305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872257 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04122574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872299 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04436318 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04429468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872276 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872253 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872266 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04436340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872247 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10522960 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522958 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522948 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522947 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522950 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522954 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522959 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522951 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522956 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522957 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522953 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522949 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522955 - -
          Affymetrix HT MG-430 PM Array Plate 1449417_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10522946 - -
          Mouse Genome 430 2.0 Array 1449417_PM_at - -
          Mouse Genome 430 2.0 Array 1449417_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11698_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11698_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500007121.mm.1 TC0500000811.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007121.mm.1 NM_009664.2 Refseq
          Mouse Expression Set 430 1449417_PM_at - -
          Mouse Expression Set 430 1449417_at - -
          Murine Genome U74v2 Set 93692_f_at - -
          Murine Genome U74v2 Set 93692_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1449417_PM_at - -
          Mouse Genome 430A 2.0 Array 1449417_at - -
          Affymetrix HT Mouse Genome 430A Array 1449417_at - -
          Affymetrix HT Mouse Genome 430A Array 1449417_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4150170 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004150170 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1240933 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1240933 - -
          mouse-6 v1.1 (Illumina) scl0011698.1_91-S - -
          Illumina Sentrix mouse-6.v1 bead array 4150170 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0011698.1_91-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4150170 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1240933 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1240933 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline