Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00477703_m1
          See other HRC GE Assays ›
          Gene Symbol:
          Hrc
          Gene Name:
          histidine rich calcium binding protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 45335269 - 45338973 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00477703_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hrc
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010473.2 NP_034603.2 4 - 5 2305 74
          GenBank mRNA AF132218.1 - 4 - 5 2284 74
          AF158597.1 - 5 - 6 2139 74
          AI322629.1 - 4 - 5 256 74
          BC141121.1 - 6 - 7 2124 9055733 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000840.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hrc

          Gene Ontology Categories:

          Function(s)

          regulation of heart contraction
          negative regulation of cytosolic calcium ion concentration
          calcium ion homeostasis
          negative regulation of calcium ion import into sarcoplasmic reticulum

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994229 AF158597.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994229 AI322629.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994229 AF132218.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994229 NM_010473.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 20776 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 670834 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 3361 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166191 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994229 AF158597.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994229 AI322629.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994229 AF132218.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994229 NM_010473.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25092506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25092500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25092503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25092512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25092487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25092492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25092491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25092488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25092481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25092511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25092501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25092485 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25092508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25092496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25092486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25092483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25092498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25092479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25092480 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25092493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25092482 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25092507 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166191 - -
          Whole Mouse Genome G4122A A_51_P267751 BC141121.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P267751 AF158597.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P267751 AI322629.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P267751 AF132218.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P267751 NM_010473.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103149 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134464 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131551 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166191 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05975399 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05975400 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05975390 - -
          Agilent Mouse cDNA Microarray (G4104A) 475063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25092482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25092475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25092487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25092492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25092491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25092507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25092483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06545852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06322776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06322774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06545847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05975395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25092477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25092478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25092476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05975385 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25092482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25092500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25092479 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25092485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25092511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25092486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25092498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25092506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25092493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25092501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06545855 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10552925 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552927 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552922 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552926 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552924 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552920 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552923 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552921 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552928 - -
          Affymetrix HT MG-430 PM Array Plate 1419109_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10552919 - -
          Mouse Genome 430 2.0 Array 1419109_at - -
          Mouse Genome 430 2.0 Array 1419109_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15464_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15464_at - -
          Affymetrix Mouse Clariom S Array PSR0700007240.mm.1 NM_010473.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700007240.mm.1 NM_010473.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700007240.mm.1 TC0700000840.mm.1 MTA TC
          Mouse Expression Set 430 1419109_PM_at - -
          Mouse Expression Set 430 1426048_s_at - -
          Mouse Expression Set 430 1419109_at - -
          Mouse Expression Set 430 1426048_PM_s_at - -
          Murine Genome U74v2 Set 92371_at - -
          Murine Genome U74v2 Set 92371_PM_at - -
          Mouse Genome 430A 2.0 Array 1419109_at - -
          Mouse Genome 430A 2.0 Array 1419109_PM_at - -
          Mouse Genome 430A 2.0 Array 1426048_s_at - -
          Mouse Genome 430A 2.0 Array 1426048_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1419109_at - -
          Affymetrix HT Mouse Genome 430A Array 1419109_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 526036 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2690563 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002690563 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1259206 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1259206 - -
          mouse-6 v1.1 (Illumina) scl32687.7.1_13-S - -
          Illumina Sentrix mouse-6.v1 bead array 2690563 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32687.7.1_13-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2690563 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1259206 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1259206 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline