Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00477734_m1
          See other PHYH GE Assays ›
          Gene Symbol:
          Phyh
          Gene Name:
          phytanoyl-CoA hydroxylase
          Gene Aliases:
          AI256161, AI265699, Lnap1, PAHX
          Chromosome Location:
          Chr.2: 4918996 - 4938743 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00477734_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Phyh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010726.2 NP_034856.1 1 - 2 151 82
          GenBank mRNA AF023463.1 - 1 - 2 116 82
          AK146753.1 - 1 - 2 136 82
          AK151372.1 - 1 - 2 101 82
          AK164495.1 - 1 - 2 102 82
          AK165412.1 - 1 - 2 95 82
          AK168648.1 - 1 - 2 121 82
          AK169213.1 - 1 - 2 93 82
          BC002018.1 - 1 - 2 128 3489939 82
          BY139974.1 - 1 - 2 151 82
          D88670.1 - 1 - 2 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000037.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Phyh

          Panther Classification:

          Molecular Function -

          hydroxylase

          Gene Ontology Categories:

          Function(s) Process(es)

          fatty acid alpha-oxidation
          isoprenoid metabolic process
          oxidation-reduction process
          methyl-branched fatty acid metabolic process
          catalytic activity
          protein binding
          oxidoreductase activity
          L-ascorbic acid binding
          metal ion binding
          cofactor binding
          phytanoyl-CoA dioxygenase activity
          dioxygenase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P276063 AF023463.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P276063 AF023463.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 447703 AK085506.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 348464 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 182267 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 76580 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 169611 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 921375 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 90563 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 302573 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 610474 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 33125 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 780552 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 164908 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 174847 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 147428 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P111235 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105360 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P276063 AF023463.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P276063 AF023463.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P276063 AF023463.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P276063 AF023463.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20970980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20970975 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20970990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20970989 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20970979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20970968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20970965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20970972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20970982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20970966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20970998 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20970994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20970995 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20970987 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20970969 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20970983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20970991 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20970985 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04414589 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P111235 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105360 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04414602 - -
          Whole Mouse Genome G4122A A_51_P276063 AF023463.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276063 AF023463.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133293 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105360 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P111235 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02244602 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02244597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20970974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20970994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20970998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20970987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20970954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20970968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20970952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20970959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20970989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20970980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20970990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20970962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20970985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20970976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20970982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20970960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20970969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971015 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01512907 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01336500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02263633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01937030 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20970983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20970968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20970982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971108 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02110196 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20970959 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01372433 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20970998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01192631 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20970992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02277375 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01388420 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20970986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01556054 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01530613 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971018 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20970980 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01250536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01192633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20970974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01643673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20970989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01372431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20970975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20970991 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02244602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20970973 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20970966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20970990 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971001 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971115 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971012 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20970994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971003 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20970962 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10469047 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469050 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469053 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469051 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469054 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469049 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469055 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469052 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469048 - -
          Affymetrix HT MG-430 PM Array Plate 1460194_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10469046 - -
          Mouse Genome 430 2.0 Array 1460194_PM_at - -
          Mouse Genome 430 2.0 Array 1460194_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16922_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16922_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200000295.mm.1 NM_010726.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000295.mm.1 NM_010726.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000295.mm.1 TC0200000037.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200000297.mm.1 NM_010726.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000297.mm.1 NM_010726.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000297.mm.1 TC0200000037.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200000296.mm.1 NM_010726.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000296.mm.1 NM_010726.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000296.mm.1 TC0200000037.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200000294.mm.1 NM_010726.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000294.mm.1 NM_010726.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000294.mm.1 TC0200000037.mm.1 MTA TC
          Mouse Expression Set 430 1460194_at - -
          Mouse Expression Set 430 1460194_PM_at - -
          Murine Genome U74v2 Set 96608_PM_at - -
          Murine Genome U74v2 Set 96608_at - -
          Mouse Genome 430A 2.0 Array 1460194_at - -
          Mouse Genome 430A 2.0 Array 1460194_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1460194_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1460194_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 416913 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6380128 - -
          Illumina mouse-6 v1.0 expression beadchip 6380128 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006380128 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2756238 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2756238 - -
          mouse-6 v1.1 (Illumina) scl21324.9.1_204-S - -
          Illumina Sentrix mouse-6.v1 bead array 6380128 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21324.9.1_204-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6380128 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2756238 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2756238 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline