Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00477804_m1
          See other NFS1 GE Assays ›
          Gene Symbol:
          Nfs1
          Gene Name:
          nitrogen fixation gene 1 (S. cerevisiae)
          Gene Aliases:
          AA987187, m-Nfs1, m-Nfsl
          Chromosome Location:
          Chr.2: 156123637 - 156144186 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00477804_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nfs1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010911.2 NP_035041.2 3 - 4 376 58
          GenBank mRNA AJ222660.1 - 3 - 4 375 58
          AK005724.1 - 3 - 4 365 58
          AK075575.1 - 3 - 4 374 58
          AK084831.1 - 3 - 4 345 58
          AK132838.1 - 3 - 4 377 58
          AK148527.1 - 3 - 4 377 58
          AK159828.1 - 3 - 4 365 58
          BC010586.1 - 3 - 4 354 3711352 58
          BC012651.1 - 3 - 4 362 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200005512.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nfs1

          Panther Classification:

          Molecular Function -

          lyase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          cysteine metabolic process
          Mo-molybdopterin cofactor biosynthetic process
          metabolic process
          iron incorporation into metallo-sulfur cluster
          catalytic activity
          transferase activity
          pyridoxal phosphate binding
          cysteine desulfurase activity
          protein homodimerization activity
          metal ion binding
          iron-sulfur cluster binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21769693 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21769694 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P315646 BC010586.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P315646 AJ222660.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P315646 BC010586.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P315646 AJ222660.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 571767 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 588884 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 839265 NM_009347.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 839265 X99805.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 217440 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 219796 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 295310 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 605481 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 9016 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 575925 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 88368 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 164203 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 596117 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 415782 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 64865 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 164184 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134222 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137506 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P315646 BC010586.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P315646 AJ222660.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P315646 BC010586.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P315646 AJ222660.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P252121 AK003786.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P252121 AK003786.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P315646 BC010586.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P315646 AJ222660.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P315646 BC010586.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P315646 AJ222660.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769674 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137506 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134222 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00534779 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00534790 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04751343 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00534772 - -
          Whole Mouse Genome G4122A A_52_P252121 AK003786.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P252121 AK003786.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P315646 BC010586.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P315646 AJ222660.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P315646 BC010586.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P315646 AJ222660.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115330 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134222 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137506 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01887019 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01887008 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01887017 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01887022 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01887006 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21769694 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21769693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01177055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01887003 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21769675 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21769693 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01151078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21769683 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21769679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21769678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21769621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01887023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21769685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01177056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21769668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01197522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01887006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01773517 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21769680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01177058 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01828879 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21769688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21769665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01351725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21769623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02169795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02159071 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10489006 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489016 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489008 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489017 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489011 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489014 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489013 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489010 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489015 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489012 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489005 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489007 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489009 - -
          Affymetrix HT MG-430 PM Array Plate 1416373_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1442017_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10489004 - -
          Mouse Genome 430 2.0 Array 1416373_at - -
          Mouse Genome 430 2.0 Array 1431431_a_at - -
          Mouse Genome 430 2.0 Array 1442017_at - -
          Mouse Genome 430 2.0 Array 1431431_PM_a_at - -
          Mouse Genome 430 2.0 Array 1442017_PM_at - -
          Mouse Genome 430 2.0 Array 1416373_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18041_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18041_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200042531.mm.1 NM_010911.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200042531.mm.1 NM_010911.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200042531.mm.1 TC0200005512.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200042528.mm.1 NM_010911.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200042528.mm.1 NM_010911.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200042528.mm.1 TC0200005512.mm.1 MTA TC
          Mouse Expression Set 430 1431431_a_at - -
          Mouse Expression Set 430 1416373_at - -
          Mouse Expression Set 430 1416373_PM_at - -
          Mouse Expression Set 430 1431431_PM_a_at - -
          Mouse Expression Set 430 1442017_at - -
          Mouse Expression Set 430 1442017_PM_at - -
          Murine Genome U74v2 Set 95065_at - -
          Murine Genome U74v2 Set 95065_PM_at - -
          Murine Genome U74v2 Set 169597_PM_at - -
          Murine Genome U74v2 Set 169597_at - -
          Mouse Genome 430A 2.0 Array 1416373_at - -
          Mouse Genome 430A 2.0 Array 1416373_PM_at - -
          Mouse Genome 430A 2.0 Array 1431431_a_at - -
          Mouse Genome 430A 2.0 Array 1431431_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1416373_at - -
          Affymetrix HT Mouse Genome 430A Array 1416373_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1442017_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1442017_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 381660 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103140056 - -
          MouseWG-6 v2 Expression BeadChip 102450369 - -
          MouseWG-6 v2 Expression BeadChip 2680563 - -
          MouseWG-6 v2 Expression BeadChip 1170465 - -
          Illumina mouse-6 v1.0 expression beadchip 103140056 - -
          Illumina mouse-6 v1.0 expression beadchip 1170465 - -
          Illumina mouse-6 v1.0 expression beadchip 2680563 - -
          Illumina mouse-6 v1.0 expression beadchip 102450369 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002680563 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001170465 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2648867 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1249158 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1235445 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1249158 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2648867 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230720 - -
          mouse-6 v1.1 (Illumina) scl0003301.1_436-S - -
          mouse-6 v1.1 (Illumina) scl18443.13.1_19-S - -
          Illumina Sentrix mouse-6.v1 bead array 103140056 - -
          Illumina Sentrix mouse-6.v1 bead array 2680563 - -
          Illumina Sentrix mouse-6.v1 bead array 102450369 - -
          Illumina Sentrix mouse-6.v1 bead array 1170465 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0003301.1_436-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl18443.13.1_19-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103140056 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2680563 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1170465 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102450369 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1249158 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2648867 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2978163 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2978163 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230720 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1249158 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1235445 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2648867 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline