Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00478870_m1
          See other PLA2G2E GE Assays ›
          Gene Symbol:
          Pla2g2e
          Gene Name:
          phospholipase A2, group IIE
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 138877911 - 138882817 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00478870_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pla2g2e
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012044.2 NP_036174.1 4 - 5 450 92
          GenBank mRNA AF112984.1 - 4 - 5 450 92
          AF166098.1 - 4 - 5 446 92
          AK148187.1 - 466 92
          BC027524.1 - 3 - 4 339 1533423 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001736.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pla2g2e

          Panther Classification:

          Molecular Function -

          phospholipase

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          phospholipid metabolic process
          lipid catabolic process
          phospholipase A2 activity
          calcium ion binding
          hydrolase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P113008 NM_012044.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P113008 AF112984.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P113008 AK148187.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P113008 BC027524.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2162807 NM_012044.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2162807 AF166098.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 412374 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 745810 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 3355 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 529554 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 946935 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 839968 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 158128 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P172950 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P113008 NM_012044.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P113008 AF112984.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P113008 AK148187.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P113008 BC027524.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2162807 NM_012044.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2162807 AF166098.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P311159 AF112984.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P311159 AK148187.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P311159 BC027524.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348409 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348406 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348416 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348394 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348414 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348398 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348387 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348390 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348380 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348413 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348403 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348384 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348400 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348412 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348411 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348386 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348425 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348382 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348408 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348420 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05423575 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P172950 - -
          Whole Mouse Genome G4122A A_51_P311159 AF112984.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311159 AK148187.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311159 BC027524.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113008 NM_012044.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113008 AF112984.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113008 AK148187.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113008 BC027524.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P172950 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03388235 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03388237 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03388245 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03388232 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03388246 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03388244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348490 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03437885 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03877608 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03476962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348399 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04074545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348448 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03255143 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04077097 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348408 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03226181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03476968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04074547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03479927 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03479924 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03437887 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03206248 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348430 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03672866 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03226190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04071767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348461 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03226184 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03226192 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03476956 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03226183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348406 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348403 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03877603 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03730031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348387 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04074546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348397 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04071766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04078322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348432 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348447 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348433 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348398 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348381 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03492763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348402 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03730040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348412 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348445 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348429 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348443 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348439 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348388 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348513 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03471952 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03205633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03476961 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348455 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03477773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03479928 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03877607 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348409 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03730032 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03476958 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10509595 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509593 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509592 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509591 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509594 - -
          Affymetrix HT MG-430 PM Array Plate 1420674_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10509590 - -
          Mouse Genome 430 2.0 Array 1420674_at - -
          Mouse Genome 430 2.0 Array 1420674_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26970_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26970_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400014179.mm.1 NM_012044.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014179.mm.1 NM_012044.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014179.mm.1 TC0400001736.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400014180.mm.1 NM_012044.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014180.mm.1 NM_012044.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014180.mm.1 TC0400001736.mm.1 MTA TC
          Mouse Expression Set 430 1421325_at - -
          Mouse Expression Set 430 1434852_at - -
          Mouse Expression Set 430 1434852_PM_at - -
          Mouse Expression Set 430 1421325_PM_at - -
          Mouse Expression Set 430 1420674_PM_at - -
          Mouse Expression Set 430 1420674_at - -
          Murine Genome U74v2 Set 165529_PM_r_at - -
          Murine Genome U74v2 Set 165529_r_at - -
          Mouse Genome 430A 2.0 Array 1434852_at - -
          Mouse Genome 430A 2.0 Array 1421325_at - -
          Mouse Genome 430A 2.0 Array 1420674_PM_at - -
          Mouse Genome 430A 2.0 Array 1421325_PM_at - -
          Mouse Genome 430A 2.0 Array 1420674_at - -
          Mouse Genome 430A 2.0 Array 1434852_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420674_at - -
          Affymetrix HT Mouse Genome 430A Array 1420674_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 505612 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4060128 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004060128 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1212812 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1212812 - -
          mouse-6 v1.1 (Illumina) scl24782.5.1_9-S - -
          Illumina Sentrix mouse-6.v1 bead array 4060128 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl24782.5.1_9-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4060128 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1212812 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1212812 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline