Search
Search

Pard6a| Gene Symbol: Pard6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047435.2 | NP_001040900.1 | 1 - 2 | 161 | 93 | |
| NM_001286345.1 | NP_001273274.1 | 1 - 2 | 161 | 93 | ||
| NM_019695.3 | NP_062669.2 | 1 - 2 | 161 | 93 | ||
| GenBank mRNA | AF070970.1 | - | 1 - 2 | 157 | 93 | |
| AK158848.1 | - | 1 - 2 | 159 | 93 | ||
| BC049593.1 | - | 1 - 2 | 138 | 6771847 | 93 | |
| BC061114.1 | - | 1 - 2 | 161 | 6535523 | 93 | |
| BG806503.1 | - | 1 - 2 | 86 | 93 | ||
| BY280124.1 | - | 1 - 2 | 159 | 93 | ||
Set Membership: |
|