Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00480135_g1
          See other PRMT1 GE Assays ›
          Gene Symbol:
          Prmt1
          Gene Name:
          protein arginine N-methyltransferase 1
          Gene Aliases:
          6720434D09Rik, AW214366, Hrmt1l2, Mrmt1
          Chromosome Location:
          Chr.7: 44976755 - 44986420 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00480135_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prmt1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001252476.1 NP_001239405.1 2 - 3 234 62
          NM_001252477.1 NP_001239406.1 2 - 3 234 62
          NM_019830.3 NP_062804.1 3 - 4 288 62
          NR_045521.1 4 - 5 433 62
          GenBank mRNA AF232716.1 - 3 - 4 200 62
          AF232717.1 - 2 - 3 146 62
          AK020120.1 - 2 - 3 153 62
          AK088790.1 - 2 - 3 146 62
          AK146927.1 - 156 62
          AK146975.1 - 2 - 3 162 62
          AK152342.1 - 2 - 3 180 62
          AK164421.1 - 2 - 3 160 62
          AK181903.1 - 2 - 3 115 62
          AK182363.1 - 2 - 3 128 62
          AK198313.1 - 2 - 3 115 62
          BC002249.1 - 2 - 3 126 3493263 62
          BC051547.1 - 2 - 3 123 62
          BC051953.1 - 4 - 5 369 5692474 62
          BC062964.1 - 4 - 5 369 5692474 62
          BU524421.1 - 3 - 4 288 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002980.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prmt1

          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          regulation of transcription, DNA-templated
          protein methylation
          histone methylation
          peptidyl-arginine methylation
          peptidyl-arginine methylation, to asymmetrical-dimethyl arginine
          neuron projection development
          methylation
          peptidyl-arginine N-methylation
          peptidyl-arginine omega-N-methylation
          histone H4-R3 methylation
          negative regulation of megakaryocyte differentiation
          rRNA (adenine-N6,N6-)-dimethyltransferase activity
          protein C-terminal carboxyl O-methyltransferase activity
          tRNA (guanine-N2-)-methyltransferase activity
          protein binding
          methyltransferase activity
          C-methyltransferase activity
          N-methyltransferase activity
          O-methyltransferase activity
          S-methyltransferase activity
          RNA methyltransferase activity
          mRNA methyltransferase activity
          tRNA methyltransferase activity
          protein methyltransferase activity
          2-polyprenyl-6-methoxy-1,4-benzoquinone methyltransferase activity
          histone-arginine N-methyltransferase activity
          rRNA methyltransferase activity
          rRNA (uridine-2'-O-)-methyltransferase activity
          rRNA (adenine-N6-)-methyltransferase activity
          DNA-methyltransferase activity
          tRNA (guanine-N1-)-methyltransferase activity
          rRNA (cytosine-C5-)-methyltransferase activity
          selenocysteine methyltransferase activity
          arginine N-methyltransferase activity
          protein-arginine N-methyltransferase activity
          [cytochrome c]-arginine N-methyltransferase activity
          lysine N-methyltransferase activity
          protein-lysine N-methyltransferase activity
          tRNA (uracil) methyltransferase activity
          tRNA (guanine) methyltransferase activity
          tRNA (adenine) methyltransferase activity
          tRNA (cytosine) methyltransferase activity
          tRNA (cytosine-5-)-methyltransferase activity
          tRNA (adenine-N1-)-methyltransferase activity
          rRNA (adenine) methyltransferase activity
          rRNA (cytosine) methyltransferase activity
          rRNA (guanine) methyltransferase activity
          rRNA (uridine) methyltransferase activity
          transferase activity
          protein C-terminal leucine carboxyl O-methyltransferase activity
          1-phenanthrol methyltransferase activity
          protein-arginine N5-methyltransferase activity
          snoRNP binding
          methylarsonite methyltransferase activity
          dimethylarsinite methyltransferase activity
          4,5-dihydroxybenzo(a)pyrene methyltransferase activity
          1-hydroxypyrene methyltransferase activity
          1-hydroxy-6-methoxypyrene methyltransferase activity
          protein-arginine omega-N monomethyltransferase activity
          protein-arginine omega-N asymmetric methyltransferase activity
          histone methyltransferase activity
          identical protein binding
          demethylmenaquinone methyltransferase activity
          cobalt-precorrin-6B C5-methyltransferase activity
          cobalt-precorrin-7 C15-methyltransferase activity
          cobalt-precorrin-5B C1-methyltransferase activity
          cobalt-precorrin-3 C17-methyltransferase activity
          dimethylamine methyltransferase activity
          hydroxyneurosporene-O-methyltransferase activity
          tRNA (adenine-57, 58-N(1)-) methyltransferase activity
          methylamine-specific methylcobalamin:coenzyme M methyltransferase activity
          trimethylamine methyltransferase activity
          methanol-specific methylcobalamin:coenzyme M methyltransferase activity
          monomethylamine methyltransferase activity
          histone methyltransferase activity (H4-R3 specific)
          poly(A) RNA binding
          P-methyltransferase activity
          Se-methyltransferase activity
          2-phytyl-1,4-naphthoquinone methyltransferase activity
          tRNA (uracil-2'-O-)-methyltransferase activity
          tRNA (cytosine-2'-O-)-methyltransferase activity
          phosphomethylethanolamine N-methyltransferase activity
          tRNA (cytosine-3-)-methyltransferase activity
          rRNA (cytosine-2'-O-)-methyltransferase activity
          rRNA (cytosine-N4-)-methyltransferase activity
          trihydroxyferuloyl spermidine O-methyltransferase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700026715.mm.1 NM_001252476.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026715.mm.1 NM_001252476.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026715.mm.1 NM_019830.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026715.mm.1 NM_019830.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026715.mm.1 NR_045521.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026715.mm.1 NR_045521.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026715.mm.1 TC0700002980.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700026716.mm.1 NM_001252476.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026716.mm.1 NM_001252476.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026716.mm.1 NM_001252477.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026716.mm.1 NM_001252477.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026716.mm.1 NM_019830.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026716.mm.1 NM_019830.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026716.mm.1 NR_045521.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026716.mm.1 NR_045521.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026716.mm.1 TC0700002980.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700026717.mm.1 NM_001252476.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026717.mm.1 NM_001252476.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026717.mm.1 NM_001252477.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026717.mm.1 NM_001252477.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026717.mm.1 NM_019830.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026717.mm.1 NM_019830.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026717.mm.1 NR_045521.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026717.mm.1 NR_045521.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026717.mm.1 TC0700002980.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700026713.mm.1 NM_001252476.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026713.mm.1 NM_001252476.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026713.mm.1 NM_019830.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026713.mm.1 NM_019830.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026713.mm.1 NR_045521.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026713.mm.1 NR_045521.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026713.mm.1 TC0700002980.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700026719.mm.1 NM_001252476.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026719.mm.1 NM_001252476.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026719.mm.1 NM_001252477.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026719.mm.1 NM_001252477.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026719.mm.1 NM_019830.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026719.mm.1 NM_019830.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026719.mm.1 NR_045521.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026719.mm.1 NR_045521.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026719.mm.1 TC0700002980.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700026718.mm.1 NM_001252476.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026718.mm.1 NM_001252476.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026718.mm.1 NM_001252477.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026718.mm.1 NM_001252477.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026718.mm.1 NM_019830.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026718.mm.1 NM_019830.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026718.mm.1 NR_045521.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026718.mm.1 NR_045521.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026718.mm.1 TC0700002980.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline