Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00480211_m1
          See other KLK11 GE Assays ›
          Gene Symbol:
          Klk11
          Gene Name:
          kallikrein related-peptidase 11
          Gene Aliases:
          2310015I08Rik, Prss20, TLSP
          Chromosome Location:
          Chr.7: 43774617 - 43779263 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00480211_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klk11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001177373.1 NP_001170844.1 3 - 4 416 68
          NM_019974.2 NP_064358.1 2 - 3 382 68
          GenBank mRNA AB016226.1 - 3 - 4 266 68
          AB016227.1 - 3 - 4 376 68
          AK009360.1 - 3 - 4 417 68
          AK009659.1 - 3 - 4 392 68
          AK009720.1 - 3 - 4 417 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000772.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protease Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klk11

          Panther Classification:

          Molecular Function -

          protease serine protease protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          serine-type endopeptidase activity
          peptidase activity
          serine-type peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979714 AK009659.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979714 AK009720.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979714 AK019102.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979714 AB016226.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979714 AB016227.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 33698 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143719 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979714 AK009659.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979714 AK009720.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979714 AK019102.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979714 AB016226.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979714 AB016227.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P154852 BC115518.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P154852 AK009659.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P154852 AK009720.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P154852 AB016226.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P154852 AB016227.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083974 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083970 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083956 - -
          Whole Mouse Genome G4122A A_51_P154852 BC115518.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154852 AK009659.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154852 AK009720.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154852 AB016226.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154852 AB016227.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06433957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06034183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06432369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06433967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06034174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06227520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06433971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083895 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10552482 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552483 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552481 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552486 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552484 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552487 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552485 - -
          Affymetrix HT MG-430 PM Array Plate 1419201_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10552480 - -
          Mouse Genome 430 2.0 Array 1419201_at - -
          Mouse Genome 430 2.0 Array 1419201_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56538_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56538_at - -
          Affymetrix Mouse Clariom S Array PSR0700006271.mm.1 TC0700000772.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700006271.mm.1 NM_001177373.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006271.mm.1 NM_001177373.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006271.mm.1 NM_019974.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006271.mm.1 NM_019974.2 Refseq
          Mouse Expression Set 430 1419201_PM_at - -
          Mouse Expression Set 430 1419201_at - -
          Murine Genome U74v2 Set 165962_PM_r_at - -
          Murine Genome U74v2 Set 165962_r_at - -
          Mouse Genome 430A 2.0 Array 1419201_at - -
          Mouse Genome 430A 2.0 Array 1419201_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419201_at - -
          Affymetrix HT Mouse Genome 430A Array 1419201_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 910400 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3360070 - -
          Illumina mouse-6 v1.0 expression beadchip 3360070 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003360070 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2718509 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2718509 - -
          mouse-6 v1.1 (Illumina) scl32743.6.27_11-S - -
          Illumina Sentrix mouse-6.v1 bead array 3360070 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32743.6.27_11-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3360070 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2780280 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2718509 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2718509 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2780280 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline