Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00481181_g1
          See other KRTCAP2 GE Assays ›
          Gene Symbol:
          Krtcap2
          Gene Name:
          keratinocyte associated protein 2
          Gene Aliases:
          0610010I12Rik, Kcp2
          Chromosome Location:
          Chr.3: 89246300 - 89249723 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00481181_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Krtcap2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025327.2 NP_079603.1 2 - 3 236 104
          GenBank mRNA AK002479.1 - 2 - 3 237 104
          AK003310.1 - 2 - 3 204 104
          BC031492.1 - 2 - 3 189 3485092 104
          BC043030.1 - 2 - 3 708 5693076 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000754.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Krtcap2

          Gene Ontology Categories:

          Function(s) Process(es)

          protein N-linked glycosylation via asparagine
          dolichyl-diphosphooligosaccharide-protein glycotransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22318141 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22318142 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P428505 AK002479.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P428505 AK003310.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P428505 NM_025327.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P428505 BC031492.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P428505 BC043030.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P428505 AK002479.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P428505 AK003310.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P428505 NM_025327.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P428505 BC031492.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P428505 BC043030.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 179596 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116004 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P428505 AK002479.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P428505 AK003310.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P428505 NM_025327.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P428505 BC031492.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P428505 BC043030.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P428505 AK002479.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P428505 AK003310.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P428505 NM_025327.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P428505 BC031492.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P428505 BC043030.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318161 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318158 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318170 - -
          Whole Mouse Genome G4122A A_51_P428505 AK002479.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P428505 AK003310.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P428505 NM_025327.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P428505 BC031492.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P428505 BC043030.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P428505 AK002479.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P428505 AK003310.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P428505 NM_025327.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P428505 BC031492.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P428505 BC043030.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127356 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119491 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02611859 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02611865 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22318141 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22318142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318148 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318156 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318170 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02663410 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03036764 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02862877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318161 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318189 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03182269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318194 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318167 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318149 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03056884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318165 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03136830 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318154 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02862878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03036766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318175 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318143 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318146 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03036752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03036760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02578708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318158 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318139 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318142 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318192 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318144 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318138 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318182 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10493488 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493489 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493486 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493485 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493487 - -
          Affymetrix HT MG-430 PM Array Plate 1417059_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1417058_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10493484 - -
          Mouse Genome 430 2.0 Array 1417058_a_at - -
          Mouse Genome 430 2.0 Array 1417058_PM_a_at - -
          Mouse Genome 430 2.0 Array 1417059_PM_at - -
          Mouse Genome 430 2.0 Array 1417059_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66059_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66059_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0300005689.mm.1 NM_025327.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005689.mm.1 TC0300000754.mm.1 MTA TC
          Mouse Expression Set 430 1417059_at - -
          Mouse Expression Set 430 1417059_PM_at - -
          Mouse Expression Set 430 1417058_PM_a_at - -
          Mouse Expression Set 430 1417058_a_at - -
          Murine Genome U74v2 Set 97268_PM_i_at - -
          Murine Genome U74v2 Set 97269_f_at - -
          Murine Genome U74v2 Set 97268_i_at - -
          Murine Genome U74v2 Set 97269_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1417059_PM_at - -
          Mouse Genome 430A 2.0 Array 1417058_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1417058_a_at - -
          Mouse Genome 430A 2.0 Array 1417059_at - -
          Affymetrix HT Mouse Genome 430A Array 1417059_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417059_at - -
          Affymetrix HT Mouse Genome 430A Array 1417058_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1417058_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 348640 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5220040 - -
          Illumina mouse-6 v1.0 expression beadchip 5220040 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005220040 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2605694 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2605694 - -
          mouse-6 v1.1 (Illumina) scl22978.7.1_18-S - -
          Illumina Sentrix mouse-6.v1 bead array 5220040 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl22978.7.1_18-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5220040 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2605694 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2605694 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl22978.7.1_18-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline