Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00481243_m1
          See other CHCHD1 GE Assays ›
          Gene Symbol:
          Chchd1
          Gene Name:
          coiled-coil-helix-coiled-coil-helix domain containing 1
          Gene Aliases:
          1110001O19Rik, 2400010G13Rik, MRP-S37
          Chromosome Location:
          Chr.14: 20703027 - 20704425 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00481243_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Chchd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025366.3 NP_079642.1 2 - 3 294 81
          GenBank mRNA AK003263.1 - 2 - 3 253 81
          AK010311.1 - 2 - 3 237 81
          AK011261.1 - 2 - 3 252 81
          AK027907.1 - 2 - 3 278 81
          AV460497.1 - 2 - 3 294 81
          BC049582.1 - 277 6772118 81
          BC052720.1 - 2 - 3 273 6818266 81
          BC064310.1 - 2 - 3 273 81
          BC092216.1 - 2 - 3 261 30609530 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400000173.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Chchd1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29448495 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29448499 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29448497 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29448501 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29448538 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29448465 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29448500 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29448498 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29448496 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29448466 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29448464 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P329251 AK011261.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P329251 AK027907.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P329251 BC049582.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P329251 BC052720.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P329251 BC064310.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P329251 AK011261.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P329251 AK027907.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P329251 BC049582.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P329251 BC052720.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P329251 BC064310.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 493283 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102245 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P329251 AK011261.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P329251 AK027907.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P329251 BC049582.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P329251 BC052720.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P329251 BC064310.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P329251 AK011261.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P329251 AK027907.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P329251 BC049582.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P329251 BC052720.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P329251 BC064310.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P329251 AK011261.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P329251 AK027907.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P329251 BC049582.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P329251 BC052720.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P329251 BC064310.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P329251 AK011261.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P329251 AK027907.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P329251 BC049582.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P329251 BC052720.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P329251 BC064310.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448454 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448448 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448461 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448466 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448462 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448457 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448474 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448455 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448472 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29448525 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102245 - -
          Whole Mouse Genome G4122A A_51_P329251 AK011261.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P329251 AK027907.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P329251 BC049582.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P329251 BC052720.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P329251 BC064310.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P329251 AK011261.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P329251 AK027907.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P329251 BC049582.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P329251 BC052720.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P329251 BC064310.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P48670 AK011261.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P48670 AK027907.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P48670 AV460497.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P48670 BC049582.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P48670 BC052720.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121473 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102245 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11529981 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11529980 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11529979 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11517581 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29448498 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29448464 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29448495 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29448496 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29448538 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29448499 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29448500 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29448501 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29448497 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29448466 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29448465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29448422 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29448455 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29448470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29448457 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29448472 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11442667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29448452 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29448468 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29448469 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29448461 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29448456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29448454 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29448448 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11442662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29448466 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29448462 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29448459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29448464 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29448450 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29448463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29448474 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10413017 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413016 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413015 - -
          Affymetrix HT MG-430 PM Array Plate 1416938_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10413014 - -
          Mouse Genome 430 2.0 Array 1416938_at - -
          Mouse Genome 430 2.0 Array 1416938_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66121_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66121_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1400001373.mm.1 NM_025366.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001373.mm.1 NM_025366.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001373.mm.1 TC1400000173.mm.1 MTA TC
          Mouse Expression Set 430 1416938_PM_at - -
          Mouse Expression Set 430 1416938_at - -
          Mouse Genome 430A 2.0 Array 1416938_at - -
          Mouse Genome 430A 2.0 Array 1416938_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416938_at - -
          Affymetrix HT Mouse Genome 430A Array 1416938_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 925081 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 110706 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000110706 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1228231 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1228231 - -
          mouse-6 v1.1 (Illumina) scl46593.3.1_27-S - -
          Illumina Sentrix mouse-6.v1 bead array 110706 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46593.3.1_27-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 110706 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1228231 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1228231 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl46593.3.1_27-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline