Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00481296_g1
          See other MED11 GE Assays ›
          Gene Symbol:
          Med11
          Gene Name:
          mediator complex subunit 11
          Gene Aliases:
          1110030J09Rik, AI465144, AW545069
          Chromosome Location:
          Chr.11: 70451931 - 70453727 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00481296_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Med11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025397.2 NP_079673.2 1 - 2 128 92
          GenBank mRNA AK003984.1 - 1 - 2 106 92
          AK008153.1 - 1 - 2 146 92
          BC025057.1 - 1 - 2 128 4238297 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000951.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Med11

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          biological_process
          protein ubiquitination
          RNA polymerase II transcription cofactor activity
          protein binding
          ubiquitin protein ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27900777 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27900779 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27900780 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27900776 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27900782 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27900778 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27900781 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349803 BC025057.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349803 NM_025397.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349803 AK003984.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349803 BC025057.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349803 NM_025397.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349803 AK003984.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 600470 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116408 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349803 BC025057.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349803 NM_025397.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349803 AK003984.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349803 BC025057.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349803 NM_025397.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349803 AK003984.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900784 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900787 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900795 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900786 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900772 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900782 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900754 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116408 - -
          Whole Mouse Genome G4122A A_51_P349803 BC025057.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349803 NM_025397.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349803 AK003984.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349803 BC025057.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349803 NM_025397.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349803 AK003984.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118898 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116408 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09119732 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09575335 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09133948 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09575340 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09575343 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09255967 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09255965 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27900781 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27900782 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27900776 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27900777 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27900780 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27900779 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27900778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09187440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09042913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09739313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09255966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09042914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900744 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900795 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09475677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09133948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09169157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09475676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09096887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09133947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09686418 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10377824 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377825 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377823 - -
          Affymetrix HT MG-430 PM Array Plate 1417739_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10377822 - -
          Mouse Genome 430 2.0 Array 1417739_at - -
          Mouse Genome 430 2.0 Array 1417739_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66172_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66172_at - -
          Affymetrix Mouse Clariom S Array PSR1100008358.mm.1 NM_025397.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008358.mm.1 NM_025397.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008358.mm.1 TC1100000951.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008361.mm.1 NM_025397.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008361.mm.1 NM_025397.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008361.mm.1 TC1100000951.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008363.mm.1 NM_025397.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008363.mm.1 NM_025397.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008363.mm.1 TC1100000951.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008362.mm.1 NM_025397.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008362.mm.1 NM_025397.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008362.mm.1 TC1100000951.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008364.mm.1 NM_025397.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008364.mm.1 NM_025397.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008364.mm.1 TC1100000951.mm.1 MTA TC
          Mouse Expression Set 430 1417739_PM_at - -
          Mouse Expression Set 430 1417739_at - -
          Murine Genome U74v2 Set 160596_at - -
          Murine Genome U74v2 Set 160596_PM_at - -
          Mouse Genome 430A 2.0 Array 1417739_PM_at - -
          Mouse Genome 430A 2.0 Array 1417739_at - -
          Affymetrix HT Mouse Genome 430A Array 1417739_at - -
          Affymetrix HT Mouse Genome 430A Array 1417739_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 471331 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6350092 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006350092 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1257061 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1257061 - -
          mouse-6 v1.1 (Illumina) scl41340.2.1_55-S - -
          Illumina Sentrix mouse-6.v1 bead array 6350092 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41340.2.1_55-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6350092 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3000536 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1257061 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1257061 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3000536 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline