Search
Search

Commd4| Gene Symbol: Commd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025417.2 | NP_079693.1 | 2 - 3 | 99 | 57 | |
| GenBank mRNA | AK004174.1 | - | 2 - 3 | 100 | 57 | |
| AK009802.1 | - | 2 - 3 | 98 | 57 | ||
| AK172194.1 | - | 1 - 2 | 1211 | 57 | ||
| BC033438.1 | - | 1 - 2 | 85 | 3995599 | 57 | |
| BC049222.1 | - | 2 - 3 | 98 | 2649359 | 57 | |
| BC055798.1 | - | 1 - 2 | 1190 | 57 | ||
Set Membership: |
|