Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00481323_m1
          See other 1110059G10RIK GE Assays ›
          Gene Symbol:
          1110059G10Rik
          Gene Name:
          RIKEN cDNA 1110059G10 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 122945089 - 122951000 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00481323_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1110059G10Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025419.4 NP_079695.2 2 - 3 338 116
          GenBank mRNA AK004312.1 - 2 - 3 334 116
          AK012115.1 - 2 - 3 334 116
          AK083316.1 - 2 - 3 311 116
          AK160617.1 - 2 - 3 339 116
          BC034133.1 - 2 - 3 296 4921846 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900003296.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 1110059G10Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26892246 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26892247 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26892248 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26892245 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P194479 BC034133.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P194479 AK083316.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P194479 AK160617.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P194479 NM_025419.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P194479 AK004312.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2131353 BC034133.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2131353 AK083316.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2131353 AK160617.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2131353 AK220157.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2131353 NM_025419.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 627890 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 660449 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 124548 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 156422 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P164308 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2131353 BC034133.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2131353 AK083316.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2131353 AK160617.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2131353 AK220157.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2131353 NM_025419.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P194479 BC034133.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P194479 AK083316.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P194479 AK160617.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P194479 NM_025419.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P194479 AK004312.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892243 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892234 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P164308 - -
          Whole Mouse Genome G4122A A_52_P26138 AK083316.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P26138 NM_025419.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P194476 BC034133.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P194476 AK083316.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P194476 AK160617.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P194476 AK220157.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P194476 NM_025419.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126634 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P164308 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07689861 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07689868 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07689865 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07689876 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26892247 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26892246 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26892245 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26892248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07689865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08001559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07689862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07511840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07773810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07572379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892204 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10597917 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597915 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597914 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597916 - -
          Affymetrix HT MG-430 PM Array Plate 1418048_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10597913 - -
          Mouse Genome 430 2.0 Array 1418048_at - -
          Mouse Genome 430 2.0 Array 1418048_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66202_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66202_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900027763.mm.1 NM_025419.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027763.mm.1 NM_025419.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027763.mm.1 TC0900003296.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900027765.mm.1 NM_025419.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027765.mm.1 NM_025419.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027765.mm.1 TC0900003296.mm.1 MTA TC
          Mouse Expression Set 430 1418048_at - -
          Mouse Expression Set 430 1418048_PM_at - -
          Mouse Genome 430A 2.0 Array 1418048_PM_at - -
          Mouse Genome 430A 2.0 Array 1418048_at - -
          Affymetrix HT Mouse Genome 430A Array 1418048_at - -
          Affymetrix HT Mouse Genome 430A Array 1418048_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 463289 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3870037 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003870037 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1250700 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1250700 - -
          mouse-6 v1.1 (Illumina) scl35180.3_165-S - -
          Illumina Sentrix mouse-6.v1 bead array 3870037 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35180.3_165-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3870037 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2825260 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2825260 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl35180.3_165-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0