Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00481336_m1
          See other RPL3L GE Assays ›
          Gene Symbol:
          Rpl3l
          Gene Name:
          ribosomal protein L3-like
          Gene Aliases:
          1110057H16Rik
          Chromosome Location:
          Chr.17: 24727829 - 24736149 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00481336_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl3l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163945.1 NP_001157417.1 7 - 8 1012 92
          NM_025425.4 NP_079701.1 4 - 5 679 92
          GenBank mRNA AK002393.1 - 4 - 5 556 92
          AK004280.1 - 4 - 5 680 92
          BC037498.1 - 4 - 5 437 92
          BC085243.1 - 4 - 5 580 92
          BC145876.1 - 4 - 5 505 40131179 92
          BC145878.1 - 4 - 5 505 40131181 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000350.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpl3l

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P243123 BC085243.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P243123 BC145876.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P243123 BC145878.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P243123 NM_001163945.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P243123 NM_025425.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P243123 BC085243.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P243123 BC145876.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P243123 BC145878.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P243123 NM_001163945.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P243123 NM_025425.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 239216 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 254825 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 539251 AK031033.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 539251 AK076655.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 215555 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 81123 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 820542 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 34120 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139905 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P243123 BC085243.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P243123 BC145876.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P243123 BC145878.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P243123 NM_001163945.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P243123 NM_025425.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P243123 BC085243.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P243123 BC145876.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P243123 BC145878.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P243123 NM_001163945.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P243123 NM_025425.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P243123 BC085243.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P243123 BC145876.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P243123 BC145878.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P243123 NM_001163945.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P243123 NM_025425.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P243123 BC085243.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P243123 BC145876.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P243123 BC145878.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P243123 NM_001163945.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P243123 NM_025425.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099266 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099275 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099269 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099271 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099279 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099268 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099265 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099282 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099287 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08740430 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139905 - -
          Whole Mouse Genome G4122A A_51_P243123 BC085243.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P243123 BC145876.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P243123 BC145878.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P243123 NM_001163945.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P243123 NM_025425.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P243123 BC085243.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P243123 BC145876.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P243123 BC145878.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P243123 NM_001163945.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P243123 NM_025425.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123087 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139905 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13415564 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13360734 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13415571 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13360732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099298 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099266 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13278925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13415561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13278928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099286 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13278923 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099258 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099287 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099275 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13425953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099249 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099278 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099279 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099257 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099273 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13237894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099263 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099265 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099269 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099282 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099271 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442591 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442587 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442589 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442593 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442595 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442588 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442592 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442594 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442586 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442590 - -
          Affymetrix HT MG-430 PM Array Plate 1449398_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10442584 - -
          Mouse Genome 430 2.0 Array 1449398_at - -
          Mouse Genome 430 2.0 Array 1449398_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66211_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66211_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700002736.mm.1 NM_001163945.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002736.mm.1 NM_001163945.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002736.mm.1 NM_025425.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002736.mm.1 NM_025425.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002736.mm.1 TC1700000350.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002737.mm.1 NM_001163945.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002737.mm.1 NM_001163945.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002737.mm.1 NM_025425.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002737.mm.1 NM_025425.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002737.mm.1 TC1700000350.mm.1 MTA TC
          Mouse Expression Set 430 1449398_PM_at - -
          Mouse Expression Set 430 1449398_at - -
          Murine Genome U74v2 Set 165907_PM_f_at - -
          Murine Genome U74v2 Set 167244_f_at - -
          Murine Genome U74v2 Set 165907_f_at - -
          Murine Genome U74v2 Set 167244_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1449398_at - -
          Mouse Genome 430A 2.0 Array 1449398_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449398_at - -
          Affymetrix HT Mouse Genome 430A Array 1449398_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 591898 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 130167 - -
          Illumina mouse-6 v1.0 expression beadchip 130167 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000130167 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2649810 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2649810 - -
          mouse-6 v1.1 (Illumina) scl51006.12.1_70-S - -
          Illumina Sentrix mouse-6.v1 bead array 130167 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl51006.12.1_70-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 130167 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2649810 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2649810 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline