Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00481395_m1
          See other TMED6 GE Assays ›
          Gene Symbol:
          Tmed6
          Gene Name:
          transmembrane emp24 protein transport domain containing 6
          Gene Aliases:
          1810015P03Rik, 1810018I24Rik
          Chromosome Location:
          Chr.8: 107061484 - 107065644 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00481395_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmed6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025458.2 NP_079734.1 3 - 4 526 89
          GenBank mRNA AK007320.1 - 3 - 4 522 89
          AK007518.1 - 3 - 4 527 89
          AK007830.1 - 3 - 4 520 89
          AK007885.1 - 3 - 4 527 89
          AK019018.1 - 1 - 2 89
          BC086923.1 - 3 - 4 526 6760985 89
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AK131840.1 - Chr. - - [-] on Build 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800002870.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmed6

          Panther Classification:

          Molecular Function -

          vesicle coat protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983528 AK007885.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983528 AK007320.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983528 AK007518.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983528 AK007830.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983528 BC086923.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983529 AK007885.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983529 AK007320.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983529 AK007518.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983529 AK007830.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983529 AK019018.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 174265 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 293659 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 145671 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101427 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983528 AK007885.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983528 AK007320.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983528 AK007518.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983528 AK007830.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983528 BC086923.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983529 AK007885.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983529 AK007320.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983529 AK007518.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983529 AK007830.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983529 AK019018.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26142908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26142923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26142931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26142907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26142912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26142903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26142934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26142936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26142927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26142920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26142929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26142900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26142943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26142938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26142917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26142906 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26142914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26142901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26142940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26142942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26142939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26142925 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101427 - -
          Whole Mouse Genome G4122A A_51_P258176 AK007885.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P258176 AK007320.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P258176 AK007518.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P258176 AK007830.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P258176 AK019018.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108618 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101427 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07156586 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07156583 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07156589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26142929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26142900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26142920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07092628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26142934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26142917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26142925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26142931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26142929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26142938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26142907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26142908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07156585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26142914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07123758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26142939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26142936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26142903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26142909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26142906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26142918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07156582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07156589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26142915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26142904 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10581520 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581521 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581519 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581522 - -
          Affymetrix HT MG-430 PM Array Plate 1416490_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10581518 - -
          Mouse Genome 430 2.0 Array 1416490_at - -
          Mouse Genome 430 2.0 Array 1416490_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66269_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66269_at - -
          Affymetrix Mouse Clariom S Array PSR0800022126.mm.1 NM_025458.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800022126.mm.1 NM_025458.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800022126.mm.1 TC0800002870.mm.1 MTA TC
          Mouse Expression Set 430 1416490_at - -
          Mouse Expression Set 430 1416490_PM_at - -
          Murine Genome U74v2 Set 165402_PM_f_at - -
          Murine Genome U74v2 Set 165402_f_at - -
          Mouse Genome 430A 2.0 Array 1416490_PM_at - -
          Mouse Genome 430A 2.0 Array 1416490_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 593530 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6510048 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006510048 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1242942 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1242942 - -
          mouse-6 v1.1 (Illumina) scl34368.4.1_11-S - -
          Illumina Sentrix mouse-6.v1 bead array 6510048 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl34368.4.1_11-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6510048 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1242942 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1242942 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline