Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00481574_g1
          See other OXLD1 GE Assays ›
          Gene Symbol:
          Oxld1
          Gene Name:
          oxidoreductase like domain containing 1
          Gene Aliases:
          1810049H13Rik
          Chromosome Location:
          Chr.11: 120456601 - 120458091 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00481574_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Oxld1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025560.2 NP_079836.1 1 - 2 253 63
          GenBank mRNA AK007842.1 - 1 - 2 254 63
          AK021150.1 - 1 - 2 241 63
          BC147291.1 - 1 - 2 246 9056342 63
          BC147292.1 - 1 - 2 246 9056341 63
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BC051593.1 - Chr. - - [-] on Build 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100004207.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Oxld1

          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28184670 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28184668 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28184669 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28184667 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P142046 AK021150.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P142046 AK007842.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P142046 NM_025560.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P142046 AK021150.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P142046 AK007842.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P142046 NM_025560.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 966033 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P135216 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P142046 AK021150.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P142046 AK007842.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P142046 NM_025560.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P142046 AK021150.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P142046 AK007842.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P142046 NM_025560.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184664 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P135216 - -
          Whole Mouse Genome G4122A A_51_P142046 AK021150.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P142046 AK007842.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P142046 NM_025560.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P142046 AK021150.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P142046 AK007842.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P142046 NM_025560.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127872 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P135216 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09224868 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09480674 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09480678 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28184668 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28184669 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28184667 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28184670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09480677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09224860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28184660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28184665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28184664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28184658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28184667 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10393803 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393802 - -
          Affymetrix HT MG-430 PM Array Plate 1460254_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1441878_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10393801 - -
          Mouse Genome 430 2.0 Array 1441878_s_at - -
          Mouse Genome 430 2.0 Array 1460254_PM_at - -
          Mouse Genome 430 2.0 Array 1460254_at - -
          Mouse Genome 430 2.0 Array 1441878_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66431_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66431_at - -
          Affymetrix Mouse Clariom S Array PSR1100040116.mm.1 NM_025560.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040116.mm.1 NM_025560.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040116.mm.1 TC1100004207.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100040115.mm.1 NM_025560.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040115.mm.1 NM_025560.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040115.mm.1 TC1100004207.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100040120.mm.1 NM_025560.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040120.mm.1 NM_025560.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040120.mm.1 TC1100004207.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100040117.mm.1 NM_025560.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040117.mm.1 NM_025560.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040117.mm.1 TC1100004207.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100040119.mm.1 NM_025560.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040119.mm.1 NM_025560.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040119.mm.1 TC1100004207.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100040118.mm.1 NM_025560.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040118.mm.1 NM_025560.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040118.mm.1 TC1100004207.mm.1 MTA TC
          Mouse Expression Set 430 1460254_PM_at - -
          Mouse Expression Set 430 1460254_at - -
          Mouse Expression Set 430 1441878_s_at - -
          Mouse Expression Set 430 1441878_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1460254_at - -
          Mouse Genome 430A 2.0 Array 1460254_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1460254_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1460254_at - -
          Affymetrix HT Mouse Genome 430B Array 1441878_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1441878_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 854659 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3290056 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003290056 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2680665 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2680665 - -
          mouse-6 v1.1 (Illumina) scl39238.2.1_49-S - -
          Illumina Sentrix mouse-6.v1 bead array 3290056 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl39238.2.1_49-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3290056 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2680665 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2680665 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl39238.2.1_49-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline