Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00481612_m1
          See other FAM213B GE Assays ›
          Gene Symbol:
          Fam213b
          Gene Name:
          family with sequence similarity 213, member B
          Gene Aliases:
          2810405K02Rik, AI836168, PM/PGFS
          Chromosome Location:
          Chr.4: 154896430 - 154899043 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00481612_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam213b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025582.3 NP_079858.2 3 - 4 359 112
          GenBank mRNA AK005188.1 - 3 - 4 360 112
          AK012999.1 - 3 - 4 358 112
          BC030453.1 - 3 - 4 353 5401948 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400004127.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fam213b

          Gene Ontology Categories:

          Function(s) Process(es)

          prostaglandin biosynthetic process
          lipid metabolic process
          fatty acid metabolic process
          fatty acid biosynthetic process
          prostaglandin metabolic process
          oxidation-reduction process
          oxidoreductase activity
          oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
          prostaglandin-F synthase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P424878 NM_025582.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P424878 AK005188.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P424878 AK012999.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P424878 BC030453.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P424878 NM_025582.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P424878 AK005188.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P424878 AK012999.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P424878 BC030453.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 919364 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 163593 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131913 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P424878 NM_025582.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P424878 AK005188.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P424878 AK012999.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P424878 BC030453.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P424878 NM_025582.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P424878 AK005188.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P424878 AK012999.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P424878 BC030453.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23434899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23434885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23434895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23434905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23434906 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23434890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23434913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23434922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23434912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23434900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23434918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23434910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23434908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23434896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23434907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23434904 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23434915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23434898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23434889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23434897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23434909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23434891 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23434902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23434887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23434883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23434919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23434893 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131913 - -
          Whole Mouse Genome G4122A A_51_P424878 NM_025582.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P424878 AK005188.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P424878 AK012999.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P424878 BC030453.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P424878 NM_025582.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P424878 AK005188.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P424878 AK012999.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P424878 BC030453.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123161 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131913 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03632478 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03632429 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03632466 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03632490 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03632450 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03632492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23434889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23434895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23434900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23434918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23434931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23434879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23434884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23434904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23434887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23434896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23434934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23434906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23434878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23434909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23434913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23434891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23434897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23434907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23434898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23434881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23434912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23434936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23434932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23434899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23434928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23434902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23434908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23434893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03632442 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03632465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03551686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03423403 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23434878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23434908 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23434916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23434902 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23434889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23434900 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23434881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23434898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23434920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23434915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03632466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23434909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23434884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23434893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23434913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03416909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03632502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23434919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23434891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23434906 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23434918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23434897 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23434895 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23434922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03632492 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23434887 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03632464 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10519057 - -
          Affymetrix Mouse Gene 1.0 ST Array 10519053 - -
          Affymetrix Mouse Gene 1.0 ST Array 10519055 - -
          Affymetrix Mouse Gene 1.0 ST Array 10519054 - -
          Affymetrix Mouse Gene 1.0 ST Array 10519058 - -
          Affymetrix Mouse Gene 1.0 ST Array 10519056 - -
          Affymetrix Mouse Gene 1.0 ST Array 10519059 - -
          Affymetrix HT MG-430 PM Array Plate 1423266_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10519052 - -
          Mouse Genome 430 2.0 Array 1423266_PM_at - -
          Mouse Genome 430 2.0 Array 1423266_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66469_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66469_at - -
          Affymetrix Mouse Clariom S Array PSR0400034502.mm.1 TC0400004127.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400034502.mm.1 NM_025582.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400034502.mm.1 NM_025582.3 Refseq
          Murine Genome U74v2 Set 171238_r_at - -
          Murine Genome U74v2 Set 171238_PM_r_at - -
          Affymetrix HT Mouse Genome 430A Array 1423266_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423266_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 450012 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4590484 - -
          Illumina mouse-6 v1.0 expression beadchip 4590484 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004590484 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1238542 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1238542 - -
          mouse-6 v1.1 (Illumina) scl066469.2_10-S - -
          Illumina Sentrix mouse-6.v1 bead array 4590484 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl066469.2_10-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4590484 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1238542 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1238542 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl066469.2_10-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline