Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00481763_m1
          See other RDM1 GE Assays ›
          Gene Symbol:
          Rdm1
          Gene Name:
          RAD52 motif 1
          Gene Aliases:
          2410008M22Rik, AW212028, Rad52b
          Chromosome Location:
          Chr.11: 101627242 - 101636091 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00481763_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rdm1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025654.2 NP_079930.1 6 - 7 770 70
          GenBank mRNA AK003135.1 - 6 - 7 766 70
          AK010457.1 - 6 - 7 771 70
          AK169668.1 - 6 - 7 1758 70
          BC026811.1 - 6 - 7 750 4237838 70
          BC055013.1 - 6 - 7 759 5135843 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001664.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rdm1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          nucleotide binding
          molecular_function
          nucleic acid binding
          DNA binding
          RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P433127 BC055013.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P433127 AK003135.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P433127 AK010457.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P433127 NM_025654.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P433127 BC026811.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P433127 BC055013.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P433127 AK003135.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P433127 AK010457.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P433127 NM_025654.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P433127 BC026811.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 890922 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 81647 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 784528 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 75041 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 294635 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 164480 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P144350 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P433127 BC055013.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P433127 AK003135.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P433127 AK010457.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P433127 NM_025654.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P433127 BC026811.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P433127 BC055013.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P433127 AK003135.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P433127 AK010457.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P433127 NM_025654.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P433127 BC026811.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28074937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28074949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28074946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28074929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28074945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28074933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28074920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28074923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28074921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28074943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28074951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28074927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28074956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28074942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28074925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28074948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28074936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28074954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28074918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28074939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28074941 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P144350 - -
          Whole Mouse Genome G4122A A_51_P433127 BC055013.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P433127 AK003135.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P433127 AK010457.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P433127 NM_025654.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P433127 BC026811.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P433127 BC055013.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P433127 AK003135.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P433127 AK010457.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P433127 NM_025654.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P433127 BC026811.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126719 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P144350 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09202204 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09268530 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09447770 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09447773 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09268525 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09447771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28075018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28075016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09268522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09351438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09057444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28075021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09300701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09116967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09695615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09447773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09497239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09562667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28075013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09136803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09514895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28075003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09554175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09116966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09428728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09417079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28075015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28074920 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10381468 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381466 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381463 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381469 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381465 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381467 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381464 - -
          Affymetrix HT MG-430 PM Array Plate 1417661_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10381462 - -
          Mouse Genome 430 2.0 Array 1417661_PM_at - -
          Mouse Genome 430 2.0 Array 1417661_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66599_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66599_at - -
          Affymetrix Mouse Clariom S Array PSR1100015674.mm.1 NM_025654.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015674.mm.1 NM_025654.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015674.mm.1 TC1100001664.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015675.mm.1 NM_025654.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015675.mm.1 NM_025654.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015675.mm.1 TC1100001664.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015673.mm.1 NM_025654.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015673.mm.1 NM_025654.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015673.mm.1 TC1100001664.mm.1 MTA TC
          Mouse Expression Set 430 1417661_at - -
          Mouse Expression Set 430 1417661_PM_at - -
          Mouse Genome 430A 2.0 Array 1417661_at - -
          Mouse Genome 430A 2.0 Array 1417661_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417661_at - -
          Affymetrix HT Mouse Genome 430A Array 1417661_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 829347 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2850487 - -
          Illumina mouse-6 v1.0 expression beadchip 2850487 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002850487 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1218934 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218934 - -
          mouse-6 v1.1 (Illumina) scl40876.7.1_108-S - -
          Illumina Sentrix mouse-6.v1 bead array 2850487 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40876.7.1_108-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2850487 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1218934 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1218934 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl40876.7.1_108-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline