| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26024830 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26024831 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26024828 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26024833 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26024829 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26024834 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2062797 |
AK169408.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2062797 |
BC027027.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2062797 |
BC040087.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2062797 |
BC050139.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2062797 |
NR_033144.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
659352 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
683121 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
280538 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
716906 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
710574 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
889344 |
AK077275.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
889344 |
AY247795.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
889344 |
AY247796.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
889344 |
BC061161.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
889344 |
D84376.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
889344 |
L43371.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
889344 |
NM_008247.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
889344 |
NM_008903.1 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
569911 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
971517 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
277026 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
520314 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
205901 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
770444 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
934541 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
633650 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
923490 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
18810 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
20278 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P169689 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P133367 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2062797 |
AK169408.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2062797 |
BC027027.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2062797 |
BC040087.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2062797 |
BC050139.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2062797 |
NR_033144.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26024828 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26024820 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26024831 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26024822 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26024836 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26024840 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26024834 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26024824 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26024826 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26024818 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26024837 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26024830 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26024823 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26024839 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P169689 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P133367 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06580757 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01785026 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P1123858 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P111003 |
AK169408.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P111003 |
BC027027.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P111003 |
BC040087.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P111003 |
BC050139.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P111003 |
NR_033144.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P373184 |
AK169408.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P373184 |
BC027027.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P373184 |
BC040087.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P373184 |
BC050139.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P373184 |
CK333687.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P126258 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P133367 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P01785026 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P169689 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07150403 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07150408 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26024834 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26024830 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26024831 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26024829 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26024833 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26024828 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024702 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024734 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024752 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024754 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024826 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024834 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024688 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024830 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024822 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024799 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024739 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024796 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024818 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024836 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024823 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024735 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024706 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024812 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024774 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024794 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024807 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024755 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024757 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024693 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024787 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024809 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024821 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024694 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024713 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024805 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024745 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024686 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024751 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024777 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024844 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024716 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024828 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024814 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024756 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024679 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024698 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024726 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024721 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024786 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024740 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024718 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024759 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024802 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024743 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024768 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024749 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024733 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024801 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024800 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024725 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024804 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024742 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024744 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024736 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024840 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024788 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024770 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024701 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024846 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024691 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024847 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024738 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024769 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024850 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26024795 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024822 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024730 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024795 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024728 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024724 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024818 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024807 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024812 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07099281 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024736 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024836 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06768053 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024823 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024824 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024838 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024732 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024796 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024733 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07150408 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024820 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024718 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024841 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024794 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024840 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024726 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06967283 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024814 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024729 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024717 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024805 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024826 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024830 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07305990 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024799 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024725 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024802 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06740737 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024721 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024804 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024821 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024790 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07119424 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024837 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024800 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024734 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024735 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024723 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024828 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26024817 |
- |
- |