Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4933425L06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025751.3 | NP_080027.1 | 1 - 2 | 232 | 96 | |
GenBank mRNA | AK016761.1 | - | 1 - 2 | 228 | 96 | |
AK016919.1 | - | 1 - 2 | 233 | 96 | ||
BC119576.1 | - | 1 - 2 | 220 | 40058060 | 96 | |
BC119577.1 | - | 1 - 2 | 220 | 40058061 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |