Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00482215_g1
          See other TMEM261 GE Assays ›
          Gene Symbol:
          Tmem261
          Gene Name:
          transmembrane protein 261
          Gene Aliases:
          1700027K24Rik, 3110001D03Rik
          Chromosome Location:
          Chr.4: 75277354 - 75278286 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00482215_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem261
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025849.3 NP_080125.1 1 - 2 308 60
          GenBank mRNA AK003640.1 - 1 - 2 279 60
          AK006432.1 - 1 - 2 235 60
          AK013937.1 - 1 - 2 311 60
          BC022621.1 - 1 - 2 332 4192355 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400002873.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem261

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23023123 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23023125 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23023126 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23023124 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P212183 NM_025849.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P212183 AK003640.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P212183 AK013937.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P212183 BC022621.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P212183 NM_025849.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P212183 AK003640.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P212183 AK013937.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P212183 BC022621.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118723 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P212183 NM_025849.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P212183 AK003640.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P212183 AK013937.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P212183 BC022621.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P212183 NM_025849.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P212183 AK003640.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P212183 AK013937.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P212183 BC022621.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023108 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023161 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023117 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023158 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23023115 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118723 - -
          Whole Mouse Genome G4122A A_51_P212183 NM_025849.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P212183 AK003640.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P212183 AK013937.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P212183 BC022621.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P212183 NM_025849.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P212183 AK003640.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P212183 AK013937.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P212183 BC022621.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P212184 NM_025849.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P212184 AK003640.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P212184 AK013937.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P212184 BC022621.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129667 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118723 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04065294 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04065287 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04065281 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04065295 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04065302 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04065289 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23023123 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23023125 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23023124 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23023126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23023129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023148 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023132 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023142 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023118 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023111 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023070 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023121 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023137 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023081 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023160 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023068 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04065294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023157 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023166 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023158 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023127 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023103 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023124 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023101 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023134 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023126 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023069 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023117 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023144 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023084 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023079 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03839183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023108 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023112 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023105 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023100 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04065289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03441516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023151 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023167 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023115 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023154 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023131 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023119 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23023140 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10513954 - -
          Affymetrix Mouse Gene 1.0 ST Array 10513953 - -
          Affymetrix HT MG-430 PM Array Plate 1423317_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10513952 - -
          Mouse Genome 430 2.0 Array 1423317_at - -
          Mouse Genome 430 2.0 Array 1423317_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66928_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66928_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400023487.mm.1 TC0400002873.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400023487.mm.1 NM_025849.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400023487.mm.1 NM_025849.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400023488.mm.1 TC0400002873.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400023488.mm.1 NM_025849.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400023488.mm.1 NM_025849.3 Refseq
          Mouse Expression Set 430 1423317_PM_at - -
          Mouse Expression Set 430 1423317_at - -
          Mouse Genome 430A 2.0 Array 1423317_PM_at - -
          Mouse Genome 430A 2.0 Array 1423317_at - -
          Affymetrix HT Mouse Genome 430A Array 1423317_at - -
          Affymetrix HT Mouse Genome 430A Array 1423317_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 908765 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5860524 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005860524 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2761260 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2761260 - -
          mouse-6 v1.1 (Illumina) scl24219.2.1_21-S - -
          Illumina Sentrix mouse-6.v1 bead array 5860524 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl24219.2.1_21-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5860524 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2971131 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2971131 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl24219.2.1_21-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline