Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00482369_m1
          See other CEP19 GE Assays ›
          Gene Symbol:
          Cep19
          Gene Name:
          centrosomal protein 19
          Gene Aliases:
          1500031L02Rik, AI428934, AL022620
          Chromosome Location:
          Chr.16: 32099802 - 32108054 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00482369_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cep19
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025892.2 NP_080168.1 3 - 4 557 79
          GenBank mRNA AK005324.1 - 3 - 4 557 79
          AK007713.1 - 3 - 4 541 79
          BC023715.1 - 2 - 3 237 5346460 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600000429.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cep19

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30650942 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30650943 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30650944 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018706 BC023715.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018706 NM_025892.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018706 AK005324.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 148330 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 32439 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 627046 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 10440 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 692961 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 55685 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149720 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018706 BC023715.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018706 NM_025892.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018706 AK005324.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30650951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30650955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30650963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30650942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30650961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30650959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30650965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30650957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30650949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30650947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30650954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30650970 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30650944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30650946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30650950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30650971 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30650968 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149720 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08553705 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08553714 - -
          Whole Mouse Genome G4122A A_51_P204730 BC023715.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P204730 NM_025892.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P204730 AK005324.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126081 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149720 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12585964 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12831987 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12721918 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12568150 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12721917 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12667737 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12568157 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12585962 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12721921 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12585966 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12585963 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12568154 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30650942 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30650944 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30650943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30651002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30651003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30651012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30650954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30650947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30650984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30650950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30650987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30650993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30651026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30650982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30650981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30650959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30651025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30651035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30650970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30651007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30651028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30651014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30650957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30651021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30651027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30651016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30650961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30650978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30650949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30650946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30650968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30651000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30651019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30651009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30650986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30650979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30650963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30650944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30650971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30650989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30651029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30650999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30651018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30651022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30650996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30650975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30650942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30650955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30650955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12516115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30651024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30650966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30651023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12595088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30650961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12550512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30651018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30650974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12901268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30650944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30650978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30651003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30650968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30650999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30651014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30650976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30650970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30651002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30651007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12831989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30650947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30650949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12550517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30650952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30650971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12667737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30650951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30650995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30650977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30651009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30650946 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30650959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30651005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12506714 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30651019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30650996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30650963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30651021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30650979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12536666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30651015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30650994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30650957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30650942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30651000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30651026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30650975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30650954 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10435006 - -
          Affymetrix Mouse Gene 1.0 ST Array 10435008 - -
          Affymetrix Mouse Gene 1.0 ST Array 10435007 - -
          Affymetrix Mouse Gene 1.0 ST Array 10435005 - -
          Affymetrix HT MG-430 PM Array Plate 1417840_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10435004 - -
          Mouse Genome 430 2.0 Array 1417840_at - -
          Mouse Genome 430 2.0 Array 1417840_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66994_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66994_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1600003981.mm.1 NM_025892.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003981.mm.1 NM_025892.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003981.mm.1 TC1600000429.mm.1 MTA TC
          Mouse Expression Set 430 1417840_PM_at - -
          Mouse Expression Set 430 1417840_at - -
          Mouse Genome 430A 2.0 Array 1417840_PM_at - -
          Mouse Genome 430A 2.0 Array 1417840_at - -
          Affymetrix HT Mouse Genome 430A Array 1417840_at - -
          Affymetrix HT Mouse Genome 430A Array 1417840_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 768564 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2760037 - -
          Illumina mouse-6 v1.0 expression beadchip 2760037 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002760037 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2666532 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2666532 - -
          mouse-6 v1.1 (Illumina) scl49245.4_458-S - -
          Illumina Sentrix mouse-6.v1 bead array 2760037 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl49245.4_458-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2760037 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2982736 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2982736 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl49245.4_458-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline