Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00482406_m1
          See other 2010109I03RIK GE Assays ›
          Gene Symbol:
          2010109I03Rik
          Gene Name:
          RIKEN cDNA 2010109I03 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 74878336 - 74881704 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00482406_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2010109I03Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025929.2 NP_080205.1 2 - 3 193 80
          GenBank mRNA AK008365.1 - 2 - 3 192 80
          AK018552.1 - 2 - 3 193 80
          AK038164.1 - 169 80
          BC116397.1 - 2 - 3 176 40101946 80
          BC116398.1 - 2 - 3 176 40101947 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001725.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 2010109I03Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P272066 AK038164.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P272066 AK008365.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P272066 AK018552.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P272066 NM_025929.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P272066 AK038164.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P272066 AK008365.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P272066 AK018552.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P272066 NM_025929.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 408587 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 671475 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 623790 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 867259 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 631427 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 902296 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 684711 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 558430 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 629315 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 345429 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 875575 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 146283 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161338 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P272066 AK038164.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P272066 AK008365.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P272066 AK018552.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P272066 NM_025929.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P272066 AK038164.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P272066 AK008365.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P272066 AK018552.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P272066 NM_025929.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30340130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30340109 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30340127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30340111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30340104 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30340134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30340115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30340118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30340103 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30340116 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30340107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30340106 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30340105 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30340129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30340099 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30340121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30340123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30340108 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30340114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30340120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30340133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30340126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30340125 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08418544 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161338 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03071757 - -
          Whole Mouse Genome G4122A A_51_P272066 AK038164.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P272066 AK008365.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P272066 AK018552.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P272066 NM_025929.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P272066 AK038164.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P272066 AK008365.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P272066 AK018552.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P272066 NM_025929.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114055 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161338 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12233751 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12233739 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12233748 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12233754 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12233733 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12233734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30340066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12277932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12023656 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340107 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12151416 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11912524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340164 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12017259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12274470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340079 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340109 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12233743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12446414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340076 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12151415 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340130 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11912532 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340100 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12023652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12151417 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12109183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12427939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30340121 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10429558 - -
          Affymetrix Mouse Gene 1.0 ST Array 10429556 - -
          Affymetrix Mouse Gene 1.0 ST Array 10429557 - -
          Affymetrix Mouse Gene 1.0 ST Array 10429559 - -
          Affymetrix HT MG-430 PM Array Plate 1449496_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10429555 - -
          Mouse Genome 430 2.0 Array 1449496_at - -
          Mouse Genome 430 2.0 Array 1449496_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67038_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67038_at - -
          Affymetrix Mouse Clariom S Array PSR1500013012.mm.1 NM_025929.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013012.mm.1 NM_025929.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013012.mm.1 TC1500001725.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500013013.mm.1 NM_025929.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013013.mm.1 NM_025929.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013013.mm.1 TC1500001725.mm.1 MTA TC
          Mouse Expression Set 430 1449496_at - -
          Mouse Expression Set 430 1449496_PM_at - -
          Murine Genome U74v2 Set 168080_PM_i_at - -
          Murine Genome U74v2 Set 168080_i_at - -
          Mouse Genome 430A 2.0 Array 1449496_PM_at - -
          Mouse Genome 430A 2.0 Array 1449496_at - -
          Affymetrix HT Mouse Genome 430A Array 1449496_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449496_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 864218 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104010164 - -
          MouseWG-6 v2 Expression BeadChip 2970132 - -
          Illumina mouse-6 v1.0 expression beadchip 2970132 - -
          Illumina mouse-6 v1.0 expression beadchip 104010164 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002970132 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2617977 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2617977 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2436984 - -
          mouse-6 v1.1 (Illumina) scl0067038.2_157-S - -
          mouse-6 v1.1 (Illumina) scl47075.4_334-S - -
          Illumina Sentrix mouse-6.v1 bead array 2970132 - -
          Illumina Sentrix mouse-6.v1 bead array 104010164 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0067038.2_157-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104010164 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2970132 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2617977 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2436984 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2617977 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline