Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00482441_m1
          See other TEX40 GE Assays ›
          Gene Symbol:
          Tex40
          Gene Name:
          testis expressed 40
          Gene Aliases:
          1700019N12Rik, A430107B04Rik, MLZ-622
          Chromosome Location:
          Chr.19: 6922426 - 6925380 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00482441_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tex40
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001039494.2 NP_001034583.1 2 - 3 394 100
          GenBank mRNA AK005644.1 - 2 - 3 373 100
          AK006134.1 - 2 - 3 397 100
          AK007089.1 - 2 - 3 370 100
          BC048452.1 - 2 - 3 331 100
          BC061075.1 - 2 - 3 336 100
          BC172038.1 - 1 - 2 551 100
          BC172042.1 - 1 - 2 551 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001021.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          ion channel transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          spermatogenesis
          biological_process
          cell differentiation
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936684 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P182257 AK006134.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P182257 AK007089.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P182257 AK020779.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P182257 BC048452.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P182257 BC061075.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P182257 AK006134.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P182257 AK007089.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P182257 AK020779.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P182257 BC048452.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P182257 BC061075.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 399695 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 776039 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160896 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P182257 AK006134.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P182257 AK007089.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P182257 AK020779.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P182257 BC048452.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P182257 BC061075.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P182257 AK006134.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P182257 AK007089.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P182257 AK020779.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P182257 BC048452.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P182257 BC061075.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P182257 AK006134.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P182257 AK007089.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P182257 AK020779.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P182257 BC048452.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P182257 BC061075.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P182257 AK006134.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P182257 AK007089.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P182257 AK020779.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P182257 BC048452.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P182257 BC061075.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936700 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936703 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936696 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160896 - -
          Whole Mouse Genome G4122A A_52_P357972 BC172042.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P357972 AK006134.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P357972 AK007089.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P357972 AK020779.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P357972 BC048452.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P182257 AK006134.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P182257 AK007089.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P182257 AK020779.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P182257 BC048452.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P182257 BC061075.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P182257 AK006134.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P182257 AK007089.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P182257 AK020779.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P182257 BC048452.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P182257 BC061075.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160896 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14340214 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14340217 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14340212 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14461896 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14461898 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14461918 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14461908 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14477785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936707 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936686 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936693 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14340217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936700 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14461898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14340226 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936704 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10465496 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465497 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465495 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465498 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465499 - -
          Affymetrix HT MG-430 PM Array Plate 1449074_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10465494 - -
          Mouse Genome 430 2.0 Array 1449074_at - -
          Mouse Genome 430 2.0 Array 1449074_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67077_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67077_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900009479.mm.1 NM_001039494.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009479.mm.1 NM_001039494.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009479.mm.1 TC1900001021.mm.1 MTA TC
          Affymetrix HT Mouse Genome 430A Array 1449074_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449074_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 301932 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1258349 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2625819 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258349 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2625819 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline