| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20293512 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20293513 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20293519 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20293520 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20293515 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20293514 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20293521 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20293518 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20293517 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2000102 |
AK033578.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2000102 |
BC026952.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2000102 |
NM_025964.3 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2083484 |
AK033578.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2083484 |
BC026952.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2083484 |
NM_025964.3 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
7762 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
126808 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
416824 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
425019 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
309246 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
798615 |
AK132106.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
798615 |
AK162283.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
617272 |
AF062476.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
617272 |
BC075657.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
617272 |
NM_009291.1 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
84247 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P173320 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P179770 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2000102 |
AK033578.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2000102 |
BC026952.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2000102 |
NM_025964.3 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2083484 |
AK033578.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2083484 |
BC026952.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2083484 |
NM_025964.3 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293520 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293553 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293525 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293491 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293497 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293517 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293493 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293498 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293506 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293551 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293538 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293511 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293524 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293540 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293512 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293522 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293523 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293496 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293545 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293502 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293526 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293505 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293495 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293529 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293500 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293550 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293514 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293534 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293509 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293541 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293544 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293527 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293531 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293547 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293542 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293554 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293503 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20293535 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P173320 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P179770 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04124960 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P493175 |
AK011433.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P140420 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P173320 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P179770 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00412446 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00412436 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00412444 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00412434 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00412432 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00412439 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00412419 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00412445 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20293512 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20293521 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20293514 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20293515 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20293517 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20293518 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20293519 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20293513 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20293520 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293498 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293525 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293436 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293517 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293432 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293457 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293509 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293573 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293455 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293484 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293453 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293468 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293495 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293464 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293505 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293561 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293528 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293559 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293481 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293524 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293473 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293514 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293488 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293511 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293538 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293553 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293466 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293523 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293562 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293534 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293440 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293477 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293429 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293439 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293535 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293463 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293544 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293506 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293472 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293576 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293572 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293442 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293541 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293532 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293550 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293564 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293542 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293438 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293446 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293479 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293556 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293474 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293569 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293520 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293458 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293512 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293497 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293451 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293502 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293557 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293475 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293470 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293443 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293450 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293526 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293500 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293489 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293461 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293546 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293503 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293496 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293460 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293527 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20293522 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293489 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293530 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293477 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293523 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293475 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293511 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293538 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293557 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293469 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293517 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293492 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293500 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293473 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293541 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293556 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293484 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293562 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293515 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293509 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00900622 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00828371 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01021371 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293553 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293550 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293495 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00882131 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00412434 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293487 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01109971 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293535 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01068318 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00412440 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293566 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293479 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293468 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293498 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293474 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293491 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293466 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01109953 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293506 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293486 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293503 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00828357 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293470 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00412437 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00783104 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00412439 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293563 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293551 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293481 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00412428 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293502 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293488 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293521 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293534 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01109962 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293555 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293493 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293548 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00828359 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293496 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00366107 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00682275 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293559 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00823594 |
- |
- |