Search
Search

Ttc14| Gene Symbol: Ttc14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290500.1 | NP_001277429.1 | 3 - 4 | 577 | 122 | |
| NM_001290502.1 | NP_001277431.1 | 3 - 4 | 815 | 122 | ||
| NM_025978.4 | NP_080254.2 | 3 - 4 | 577 | 122 | ||
| NM_027619.4 | NP_081895.2 | 3 - 4 | 577 | 122 | ||
| GenBank mRNA | AK012246.1 | - | 3 - 4 | 539 | 122 | |
| AK015308.1 | - | 3 - 4 | 537 | 122 | ||
| AK016619.1 | - | 3 - 4 | 497 | 122 | ||
| AK017763.1 | - | 1 - 2 | 175 | 122 | ||
| AK031605.1 | - | 3 - 4 | 808 | 122 | ||
| AK035570.1 | - | 3 - 4 | 547 | 122 | ||
| AK041095.1 | - | 3 - 4 | 551 | 122 | ||
| AK044266.1 | - | 3 - 4 | 552 | 122 | ||
| AK134539.1 | - | 3 - 4 | 566 | 122 | ||
| AK173315.1 | - | 3 - 4 | 547 | 122 | ||
| BC021448.1 | - | 3 - 4 | 585 | 122 | ||
| BC024847.1 | - | 3 - 4 | 542 | 5355806 | 122 | |
| BC138212.1 | - | 3 - 4 | 539 | 8861231 | 122 | |
| BC145944.1 | - | 3 - 4 | 539 | 40131247 | 122 | |
Set Membership: |
|