Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00482712_m1
          See other LYZL1 GE Assays ›
          Gene Symbol:
          Lyzl1
          Gene Name:
          lysozyme-like 1
          Gene Aliases:
          1700038F02Rik, Lyc2, Lyzd1
          Chromosome Location:
          Chr.18: 4165832 - 4182236 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00482712_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lyzl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026092.3 NP_080368.1 3 - 4 442 63
          GenBank mRNA AK006241.1 - 3 - 4 443 63
          AK006630.1 - 3 - 4 443 63
          AY572447.1 - 3 - 4 401 63
          BC048450.1 - 3 - 4 413 6774504 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000011.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lyzl1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          metabolic process
          molecular_function
          lysozyme activity
          hydrolase activity
          hydrolase activity, acting on glycosyl bonds

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31461138 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31461136 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31461137 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152696 NM_026092.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152696 AK006241.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152696 AK006630.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152696 AY572447.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152696 BC048450.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152699 NM_026092.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152699 AK006630.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152699 BC048450.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 707479 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 521868 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 176084 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 950913 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 910814 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 818305 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 228465 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 172503 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165443 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P129130 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152699 NM_026092.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152699 AK006630.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152699 BC048450.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152696 NM_026092.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152696 AK006241.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152696 AK006630.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152696 AY572447.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152696 BC048450.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P172935 NM_026092.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P172935 AK006241.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P172935 AK006630.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P172935 AY572447.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P172935 BC048450.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31461778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31461779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31461789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31461781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31461782 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31461795 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31461787 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31461793 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31461785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31461776 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P129130 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165443 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08897799 - -
          Whole Mouse Genome G4122A A_51_P172935 NM_026092.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P172935 AK006241.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P172935 AK006630.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P172935 AY572447.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P172935 BC048450.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132154 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08897799 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P129130 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165443 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13844878 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13948135 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13844876 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31461138 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31461136 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31461137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31461908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13809540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461813 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13809511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461779 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461773 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13851425 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13851429 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461778 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13992078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13809534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13851430 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13809532 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13844876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13923713 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13809522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13887062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461772 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14093681 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13851420 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13809528 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461805 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31461809 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10453614 - -
          Affymetrix Mouse Gene 1.0 ST Array 10453611 - -
          Affymetrix Mouse Gene 1.0 ST Array 10453610 - -
          Affymetrix Mouse Gene 1.0 ST Array 10453612 - -
          Affymetrix Mouse Gene 1.0 ST Array 10453613 - -
          Affymetrix Mouse Gene 1.0 ST Array 10453615 - -
          Affymetrix HT MG-430 PM Array Plate 1449892_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10453609 - -
          Mouse Genome 430 2.0 Array 1449892_PM_at - -
          Mouse Genome 430 2.0 Array 1449892_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67328_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67328_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1800000072.mm.1 NM_026092.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800000072.mm.1 NM_026092.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800000072.mm.1 TC1800000011.mm.1 MTA TC
          Mouse Expression Set 430 1449892_PM_at - -
          Mouse Expression Set 430 1449892_at - -
          Mouse Genome 430A 2.0 Array 1449892_at - -
          Mouse Genome 430A 2.0 Array 1449892_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449892_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449892_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 319819 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6550324 - -
          Illumina mouse-6 v1.0 expression beadchip 6550324 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006550324 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2618942 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2618942 - -
          mouse-6 v1.1 (Illumina) scl52301.4.1_32-S - -
          Illumina Sentrix mouse-6.v1 bead array 6550324 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52301.4.1_32-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6550324 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2618942 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2618942 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline