Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00482745_m1
          See other 1700093K21RIK GE Assays ›
          Gene Symbol:
          1700093K21Rik
          Gene Name:
          RIKEN cDNA 1700093K21 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 23516203 - 23521317 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00482745_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700093K21Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001110133.1 NP_001103603.1 4 - 5 915 70
          NM_026105.3 NP_080381.2 4 - 5 597 70
          GenBank mRNA AK006386.1 - 1 - 2 101 70
          AK007052.1 - 4 - 5 70
          AK007139.1 - 3 - 4 354 70
          BC049567.1 - 4 - 5 597 6773879 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002366.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 1700093K21Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P326550 NM_026105.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P326550 AK007052.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P326550 BC049567.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P326550 NM_001110133.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P326550 NM_026105.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P326550 AK007052.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P326550 BC049567.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P326550 NM_001110133.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 354016 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 728987 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 724395 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 232290 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 342189 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 782528 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 717886 NM_001099299.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 57830 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 116407 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 84545 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 86904 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 101543 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126372 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P114884 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107277 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P326550 NM_026105.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P326550 AK007052.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P326550 BC049567.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P326550 NM_001110133.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P326550 NM_026105.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P326550 AK007052.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P326550 BC049567.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P326550 NM_001110133.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27646266 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27646262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27646264 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27646261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27646278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27646280 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27646271 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27646267 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27646272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27646279 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27646281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27646269 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27646276 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27646274 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27646260 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P114884 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126372 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107277 - -
          Whole Mouse Genome G4122A A_51_P326550 NM_026105.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P326550 AK007052.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P326550 BC049567.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P326550 NM_001110133.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P326550 NM_026105.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P326550 AK007052.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P326550 BC049567.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P326550 NM_001110133.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P326555 AK007052.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137550 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P114884 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107277 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126372 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09272191 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09272195 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09272197 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09272192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27646234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27646258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27646228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27646268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27646288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27646260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27646251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27646257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27646265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27646240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27646244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27646291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27646272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27646246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27646280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27646262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27646279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27646293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27646289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27646252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27646276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27646249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27646269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27646264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27646266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27646274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09403780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09440576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09325214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646288 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09013797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27646252 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10384687 - -
          Affymetrix Mouse Gene 1.0 ST Array 10384688 - -
          Affymetrix Mouse Gene 1.0 ST Array 10384689 - -
          Affymetrix Mouse Gene 1.0 ST Array 10384686 - -
          Affymetrix Mouse Gene 1.0 ST Array 10384690 - -
          Affymetrix HT MG-430 PM Array Plate 1420702_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10384685 - -
          Mouse Genome 430 2.0 Array 1420702_at - -
          Mouse Genome 430 2.0 Array 1420702_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67358_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67358_at - -
          Affymetrix Mouse Clariom S Array PSR1100022298.mm.1 NM_001110133.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100022298.mm.1 NM_001110133.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100022298.mm.1 NM_026105.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100022298.mm.1 NM_026105.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100022298.mm.1 TC1100002366.mm.1 MTA TC
          Mouse Expression Set 430 1420702_at - -
          Mouse Expression Set 430 1420702_PM_at - -
          Murine Genome U74v2 Set 168242_i_at - -
          Murine Genome U74v2 Set 168242_PM_i_at - -
          Mouse Genome 430A 2.0 Array 1420702_PM_at - -
          Mouse Genome 430A 2.0 Array 1420702_at - -
          Affymetrix HT Mouse Genome 430A Array 1420702_at - -
          Affymetrix HT Mouse Genome 430A Array 1420702_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 696891 - -
          ABI Mouse Genome Survey Microarray 749960 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 450551 - -
          Illumina mouse-6 v1.0 expression beadchip 450551 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000450551 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2736976 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2736976 - -
          mouse-6 v1.1 (Illumina) scl40443.6.1_30-S - -
          Illumina Sentrix mouse-6.v1 bead array 450551 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40443.6.1_30-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 450551 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2736976 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2736976 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl40443.6.1_30-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline