Search
Search
| Gene Symbol: Cbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007623.2 | NP_031649.2 | 2 - 3 | 193 | 80 | |
| GenBank mRNA | AK157869.1 | - | 2 - 3 | 169 | 80 | |
| BC035199.1 | - | 2 - 3 | 193 | 5137505 | 80 | |
| X62537.1 | - | 2 - 3 | 260 | 80 | ||
| Y13274.1 | - | 2 - 3 | 117 | 80 | ||
Set Membership: |
|