Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00483111_m1
          See other CCT5 GE Assays ›
          Gene Symbol:
          Cct5
          Gene Name:
          chaperonin containing Tcp1, subunit 5 (epsilon)
          Gene Aliases:
          Ccte, TCPE, mKIAA0098
          Chromosome Location:
          Chr.15: 31590813 - 31601804 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00483111_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cct5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007637.2 NP_031663.1 3 - 4 432 98
          GenBank mRNA AK077753.1 - 3 - 4 390 98
          AK088184.1 - 3 - 4 366 98
          AK129056.1 - 3 - 4 338 98
          AK133857.1 - 3 - 4 390 98
          AK136493.1 - 3 - 4 384 98
          AK145445.1 - 3 - 4 401 98
          AK146425.1 - 3 - 4 384 98
          AK150390.1 - 3 - 4 351 98
          AK153911.1 - 3 - 4 408 98
          AK167895.1 - 3 - 4 373 98
          AK217822.1 - 2 - 3 185 98
          BC092233.1 - 3 - 4 402 98
          BC094427.1 - 3 - 4 379 30537966 98
          Z31555.1 - 3 - 4 432 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001326.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cct5

          Panther Classification:

          Molecular Function -

          chaperonin

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          binding of sperm to zona pellucida
          toxin transport
          nucleotide binding
          protein binding
          ATP binding
          G-protein beta-subunit binding
          beta-tubulin binding
          unfolded protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30115244 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30115245 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30115243 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30115241 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30115247 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30115242 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30115248 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30115249 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30115246 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144783 BC092233.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144783 BC094427.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144783 Z31555.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144783 AK150390.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144783 AK167895.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144783 BC092233.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144783 BC094427.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144783 Z31555.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144783 AK150390.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144783 AK167895.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 18443 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 191300 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 309453 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 821856 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 898394 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 425640 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 159813 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109516 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P178219 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144783 BC092233.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144783 BC094427.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144783 Z31555.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144783 AK150390.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144783 AK167895.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144783 BC092233.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144783 BC094427.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144783 Z31555.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144783 AK150390.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144783 AK167895.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P144783 BC092233.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P144783 BC094427.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P144783 Z31555.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P144783 AK150390.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P144783 AK167895.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P144783 BC092233.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P144783 BC094427.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P144783 Z31555.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P144783 AK150390.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P144783 AK167895.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115247 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115225 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P109516 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P178219 - -
          Whole Mouse Genome G4122A A_51_P144783 BC092233.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144783 BC094427.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144783 Z31555.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144783 AK150390.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144783 AK167895.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144783 BC092233.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144783 BC094427.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144783 Z31555.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144783 AK150390.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144783 AK167895.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139669 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106014 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P178219 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P109516 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11853836 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11853827 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11853832 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11853823 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30115244 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30115243 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30115241 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30115246 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30115249 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30115242 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30115245 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30115248 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30115247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115204 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30115231 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12159146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30115242 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30115225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30115227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30115220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12020513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30115198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30115217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30115196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30115201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30115199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12159151 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12096296 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30115207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30115234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30115235 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11853829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30115212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30115247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30115197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30115219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30115246 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30115238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30115210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30115228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30115209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30115221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30115216 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30115240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30115230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30115214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12096289 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10428057 - -
          Affymetrix Mouse Gene 1.0 ST Array 10428062 - -
          Affymetrix Mouse Gene 1.0 ST Array 10428065 - -
          Affymetrix Mouse Gene 1.0 ST Array 10428058 - -
          Affymetrix Mouse Gene 1.0 ST Array 10428059 - -
          Affymetrix Mouse Gene 1.0 ST Array 10428063 - -
          Affymetrix Mouse Gene 1.0 ST Array 10428064 - -
          Affymetrix Mouse Gene 1.0 ST Array 10428054 - -
          Affymetrix Mouse Gene 1.0 ST Array 10428056 - -
          Affymetrix Mouse Gene 1.0 ST Array 10428060 - -
          Affymetrix Mouse Gene 1.0 ST Array 10428061 - -
          Affymetrix Mouse Gene 1.0 ST Array 10428055 - -
          Affymetrix HT MG-430 PM Array Plate 1417258_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10428052 - -
          Mouse Genome 430 2.0 Array 1417258_at - -
          Mouse Genome 430 2.0 Array 1417258_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12465_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12465_at - -
          Affymetrix Mouse Clariom S Array PSR1500010301.mm.1 NM_007637.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500010301.mm.1 TC1500001326.mm.1 MTA TC
          Mouse Expression Set 430 1417258_at - -
          Mouse Expression Set 430 1417258_PM_at - -
          Murine Genome U74v2 Set 93295_at - -
          Murine Genome U74v2 Set 161514_PM_at - -
          Murine Genome U74v2 Set 161514_at - -
          Murine Genome U74v2 Set 93295_PM_at - -
          Mouse Genome 430A 2.0 Array 1417258_at - -
          Mouse Genome 430A 2.0 Array 1417258_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417258_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417258_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 576286 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1050670 - -
          Illumina mouse-6 v1.0 expression beadchip 1050670 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001050670 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2707308 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2707308 - -
          mouse-6 v1.1 (Illumina) scl47326.13.2_27-S - -
          Illumina Sentrix mouse-6.v1 bead array 1050670 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl47326.13.2_27-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1050670 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2707308 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2707308 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl47326.13.2_27-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline