Search
Search

Col1a2| Gene Symbol: Col1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007743.2 | NP_031769.2 | 1 - 2 | 210 | 67 | |
| GenBank mRNA | AK031577.1 | - | 1 - 2 | 206 | 67 | |
| AK132496.1 | - | 1 - 2 | 206 | 67 | ||
| AK159212.1 | - | 1 - 2 | 211 | 67 | ||
| AK159408.1 | - | 1 - 2 | 207 | 67 | ||
| AK159578.1 | - | 1 - 2 | 207 | 67 | ||
| AK160098.1 | - | 1 - 2 | 210 | 67 | ||
| AK160787.1 | - | 1 - 2 | 206 | 67 | ||
| AK160788.1 | - | 1 - 2 | 205 | 67 | ||
| AK160790.1 | - | 1 - 2 | 206 | 67 | ||
| AK160946.1 | - | 1 - 2 | 209 | 67 | ||
| AK161017.1 | - | 1 - 2 | 211 | 67 | ||
| AK161605.1 | - | 1 - 2 | 206 | 67 | ||
| AK164808.1 | - | 1 - 2 | 206 | 67 | ||
| BC007158.1 | - | 1 - 2 | 190 | 3487477 | 67 | |
| BC042503.1 | - | 1 - 2 | 198 | 3586272 | 67 | |
| X58251.1 | - | 1 - 2 | 146 | 67 | ||
Set Membership: |
|