Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00484484_s1
          See other FPR-RS4 GE Assays ›
          Gene Symbol:
          Fpr-rs4
          Gene Name:
          formyl peptide receptor, related sequence 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 18021733 - 18022704 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00484484_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fpr-rs4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008041.2 NP_032067.2 1 842 90
          GenBank mRNA BC115534.1 - 1 838 90
          BC115535.1 - 1 842 90

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fpr-rs4

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          signal transducer activity
          G-protein coupled receptor activity
          N-formyl peptide receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1996896 BC115534.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1996896 BC115535.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1996896 NM_008041.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996896 BC115534.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996896 BC115535.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996896 NM_008041.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31075808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31075816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31075829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31075812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31075825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31075830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31075845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31075833 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31075842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31075844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31075824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31075827 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31075803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31075823 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31075832 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31075849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31075813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31075806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31075818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31075854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31075834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31075852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31075821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31075836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31075846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31075801 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31075810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31075814 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102429 - -
          Whole Mouse Genome G4122A A_52_P58424 BC115534.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58424 BC115535.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58424 NM_008041.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122305 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102429 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442105 - -
          Affymetrix HT MG-430 PM Array Plate 1422358_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10442104 - -
          Mouse Genome 430 2.0 Array 1422358_at - -
          Mouse Genome 430 2.0 Array 1422358_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14291_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14291_at - -
          Mouse Expression Set 430 1422358_at - -
          Mouse Expression Set 430 1422358_PM_at - -
          Murine Genome U74v2 Set 101802_PM_at - -
          Murine Genome U74v2 Set 101802_at - -
          Mouse Genome 430A 2.0 Array 1422358_at - -
          Mouse Genome 430A 2.0 Array 1422358_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422358_at - -
          Affymetrix HT Mouse Genome 430A Array 1422358_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 616579 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2030082 - -
          Illumina mouse-6 v1.0 expression beadchip 2030082 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002030082 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2596629 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2596629 - -
          mouse-6 v1.1 (Illumina) scl8648.1.1_181-S - -
          Illumina Sentrix mouse-6.v1 bead array 2030082 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl8648.1.1_181-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2030082 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2596629 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline