| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900001337.mm.1 |
NM_001168488.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001337.mm.1 |
NM_001168488.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001337.mm.1 |
TC1900000113.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900001346.mm.1 |
NM_001168490.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001346.mm.1 |
NM_001168490.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001346.mm.1 |
NM_008583.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001346.mm.1 |
NM_008583.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001346.mm.1 |
TC1900000113.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900001344.mm.1 |
NM_001168490.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001344.mm.1 |
NM_001168490.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001344.mm.1 |
NM_008583.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001344.mm.1 |
NM_008583.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001344.mm.1 |
TC1900000113.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900001352.mm.1 |
NM_001168488.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001352.mm.1 |
NM_001168488.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001352.mm.1 |
NM_001168489.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001352.mm.1 |
NM_001168489.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001352.mm.1 |
NM_001168490.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001352.mm.1 |
NM_001168490.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001352.mm.1 |
NM_008583.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001352.mm.1 |
NM_008583.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001352.mm.1 |
TC1900000113.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900001353.mm.1 |
NM_001168488.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001353.mm.1 |
NM_001168488.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001353.mm.1 |
NM_001168489.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001353.mm.1 |
NM_001168489.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001353.mm.1 |
NM_001168490.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001353.mm.1 |
NM_001168490.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001353.mm.1 |
NM_008583.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001353.mm.1 |
NM_008583.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001353.mm.1 |
TC1900000113.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900001347.mm.1 |
NM_001168490.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001347.mm.1 |
NM_001168490.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001347.mm.1 |
NM_008583.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001347.mm.1 |
NM_008583.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001347.mm.1 |
TC1900000113.mm.1 |
MTA TC |