Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00485703_m1
          See other RFNG GE Assays ›
          Gene Symbol:
          Rfng
          Gene Name:
          RFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 120780745 - 120784217 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00485703_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rfng
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009053.2 NP_033079.1 6 - 7 869 85
          GenBank mRNA AF015770.1 - 6 - 7 827 85
          AK004573.1 - 4 - 5 1221 85
          AK178490.1 - 1 - 2 134 86
          AK189194.1 - 1 - 2 134 85
          BC066023.1 - 6 - 7 869 6829176 85
          U94350.1 - 6 - 7 862 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100004231.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rfng

          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          pattern specification process
          nervous system development
          cell differentiation
          positive regulation of protein binding
          negative regulation of Notch signaling pathway
          positive regulation of Notch signaling pathway
          transferase activity
          transferase activity, transferring glycosyl groups
          O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28186728 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P230552 U94350.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P230552 U94350.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2109429 BC066023.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2109429 NM_009053.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2109429 AK004573.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 707973 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 75665 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123967 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2109429 BC066023.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2109429 NM_009053.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2109429 AK004573.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P230552 U94350.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P230552 U94350.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P230552 U94350.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P230552 U94350.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186709 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186712 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186717 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186723 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186726 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186724 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186728 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186708 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186729 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28186725 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123967 - -
          Whole Mouse Genome G4122A A_51_P230552 U94350.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P230552 U94350.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127790 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123967 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08992552 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28186728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28186681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09540896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08992559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09129044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08992552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09452023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09452027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09569178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09452024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186722 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09129042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28186681 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10393946 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393945 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393947 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393950 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393953 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393949 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393948 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393952 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393954 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393951 - -
          Affymetrix HT MG-430 PM Array Plate 1460704_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1421160_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10393944 - -
          Mouse Genome 430 2.0 Array 1460704_at - -
          Mouse Genome 430 2.0 Array 1421160_a_at - -
          Mouse Genome 430 2.0 Array 1421160_PM_a_at - -
          Mouse Genome 430 2.0 Array 1460704_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19719_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19719_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100040485.mm.1 NM_009053.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040485.mm.1 NM_009053.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040485.mm.1 TC1100004231.mm.1 MTA TC
          Mouse Expression Set 430 1421160_PM_a_at - -
          Mouse Expression Set 430 1460704_PM_at - -
          Mouse Expression Set 430 1460704_at - -
          Mouse Expression Set 430 1421160_a_at - -
          Murine Genome U74v2 Set 101858_PM_at - -
          Murine Genome U74v2 Set 101858_at - -
          Mouse Genome 430A 2.0 Array 1460704_at - -
          Mouse Genome 430A 2.0 Array 1421160_a_at - -
          Mouse Genome 430A 2.0 Array 1421160_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1460704_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1460704_at - -
          Affymetrix HT Mouse Genome 430A Array 1460704_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421160_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1421160_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 374901 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6290086 - -
          MouseWG-6 v2 Expression BeadChip 104780484 - -
          Illumina mouse-6 v1.0 expression beadchip 104780484 - -
          Illumina mouse-6 v1.0 expression beadchip 6290086 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006290086 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1257829 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1257829 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2699250 - -
          mouse-6 v1.1 (Illumina) scl0019719.2_122-S - -
          mouse-6 v1.1 (Illumina) scl39221.6_560-S - -
          Illumina Sentrix mouse-6.v1 bead array 6290086 - -
          Illumina Sentrix mouse-6.v1 bead array 104780484 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0019719.2_122-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104780484 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6290086 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1257829 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1257829 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0019719.2_122-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline