Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00485707_m1
          See other SLC50A1 GE Assays ›
          Gene Symbol:
          Slc50a1
          Gene Name:
          solute carrier family 50 (sugar transporter), member 1
          Gene Aliases:
          MmSWEET1, Rag1ap1, Rga
          Chromosome Location:
          Chr.3: 89268246 - 89270570 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00485707_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc50a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009057.3 NP_033083.2 3 - 4 384 111
          GenBank mRNA AK014305.1 - 3 - 4 337 111
          AK045924.1 - 1 - 2 697 111
          AK079716.1 - 2 - 3 449 111
          AK158862.1 - 2 - 3 409 111
          AK170162.1 - 3 - 4 335 111
          AK177059.1 - 3 - 4 292 111
          BC014292.1 - 3 - 4 368 4190124 111
          BY333933.1 - 3 - 4 255 111
          X96618.1 - 3 - 4 375 111

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300002343.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc50a1

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA recombination
          transport
          carbohydrate transport
          glucoside transport
          positive regulation of gene expression, epigenetic
          transmembrane transport
          glucoside transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000603 AK170162.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000603 BC014292.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000603 X96618.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000603 AK014305.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000603 NM_009057.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2069885 AK170162.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2069885 BC014292.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2069885 X96618.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2069885 AK014305.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2069885 AK045924.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 115753 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 511821 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119686 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2069885 AK170162.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2069885 BC014292.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2069885 X96618.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2069885 AK014305.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2069885 AK045924.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000603 AK170162.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000603 BC014292.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000603 X96618.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000603 AK014305.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000603 NM_009057.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318264 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318271 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318268 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318270 - -
          Whole Mouse Genome G4122A A_52_P658513 AK170162.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P658513 BC014292.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P658513 X96618.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P658513 AK014305.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P658513 AK045924.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136665 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03031842 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03031821 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03031845 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03031830 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03031851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318246 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02498909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318243 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03031848 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318256 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03031828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318244 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318271 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10499534 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499531 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499535 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499532 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499530 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499533 - -
          Affymetrix HT MG-430 PM Array Plate 1448948_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10499529 - -
          Mouse Genome 430 2.0 Array 1448948_at - -
          Mouse Genome 430 2.0 Array 1448948_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19729_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19729_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0300017948.mm.1 NM_009057.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017948.mm.1 NM_009057.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017948.mm.1 TC0300002343.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300017949.mm.1 NM_009057.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017949.mm.1 NM_009057.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017949.mm.1 TC0300002343.mm.1 MTA TC
          Mouse Expression Set 430 1448948_at - -
          Mouse Expression Set 430 1448948_PM_at - -
          Murine Genome U74v2 Set 102960_at - -
          Murine Genome U74v2 Set 102960_PM_at - -
          Mouse Genome 430A 2.0 Array 1448948_at - -
          Mouse Genome 430A 2.0 Array 1448948_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448948_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448948_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 922964 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1450008 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001450008 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2614486 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2614486 - -
          mouse-6 v1.1 (Illumina) scl21948.4.1_81-S - -
          Illumina Sentrix mouse-6.v1 bead array 1450008 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21948.4.1_81-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1450008 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2614486 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2614486 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl21948.4.1_81-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline