Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00486494_m1
          See other SURF6 GE Assays ›
          Gene Symbol:
          Surf6
          Gene Name:
          surfeit gene 6
          Gene Aliases:
          D2Wsu129e, Surf-6
          Chromosome Location:
          Chr.2: 26890772 - 26902813 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00486494_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Surf6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009298.3 NP_033324.1 1 - 2 118 58
          GenBank mRNA AK077989.1 - 1 - 2 120 58
          AK132194.1 - 1 - 2 154 58
          BC037634.1 - 1 - 2 165 4952465 58
          X92842.1 - 1 - 2 101 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003202.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Surf6

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          ribosome biogenesis
          ribosome assembly
          DNA binding
          RNA binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21081013 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21081015 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21081016 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21081014 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974317 BC037634.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974317 BC026514.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974317 AK132194.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974317 AK075636.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974317 AK077989.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010962 X92842.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010962 BC037634.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010962 BC026514.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010962 AK132194.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010962 AK077989.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463081 AK017702.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 568892 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 370528 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 794132 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 203063 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 406519 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 282277 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 823897 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 20818 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136613 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974317 BC037634.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974317 BC026514.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974317 AK132194.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974317 AK075636.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974317 AK077989.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010962 X92842.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010962 BC037634.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010962 BC026514.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010962 AK132194.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010962 AK077989.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21081030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21081020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21081015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21081014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21081006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21081019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21081017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21081029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21081011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21081012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21081023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21081021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21081027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21081009 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04461450 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136613 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04461469 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04461473 - -
          Whole Mouse Genome G4122A A_51_P311965 X92842.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311965 BC037634.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311965 BC026514.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311965 AK077989.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311965 NM_009298.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P283119 AK164904.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133160 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136613 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01348211 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01348187 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01348208 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01348190 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01348198 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21081014 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21081016 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21081015 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21081013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21081089 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21081003 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01359622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21081009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21080998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21081020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21081012 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21081030 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21081007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01866847 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01348190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01348216 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01348207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21081000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21081017 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21081029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21081023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01866852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01866850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21081010 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21081011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21081021 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10481126 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481125 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481127 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481124 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481123 - -
          Affymetrix HT MG-430 PM Array Plate 1459331_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1416864_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10481122 - -
          Mouse Genome 430 2.0 Array 1459331_at - -
          Mouse Genome 430 2.0 Array 1459331_PM_at - -
          Mouse Genome 430 2.0 Array 1416864_at - -
          Mouse Genome 430 2.0 Array 1416864_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20935_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20935_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200026894.mm.1 NM_009298.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026894.mm.1 TC0200003202.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200026896.mm.1 NM_009298.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026896.mm.1 TC0200003202.mm.1 MTA TC
          Mouse Expression Set 430 1416864_at - -
          Mouse Expression Set 430 1416864_PM_at - -
          Mouse Expression Set 430 1459331_PM_at - -
          Mouse Expression Set 430 1459331_at - -
          Murine Genome U74v2 Set 96663_at - -
          Murine Genome U74v2 Set 96663_PM_at - -
          Mouse Genome 430A 2.0 Array 1416864_PM_at - -
          Mouse Genome 430A 2.0 Array 1416864_at - -
          Affymetrix HT Mouse Genome 430A Array 1416864_at - -
          Affymetrix HT Mouse Genome 430A Array 1416864_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1459331_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1459331_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 859645 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104010440 - -
          MouseWG-6 v2 Expression BeadChip 102510176 - -
          Illumina mouse-6 v1.0 expression beadchip 3060451 - -
          Illumina mouse-6 v1.0 expression beadchip 104010440 - -
          Illumina mouse-6 v1.0 expression beadchip 102510176 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003060451 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2753108 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1233178 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2753108 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222350 - -
          mouse-6 v1.1 (Illumina) scl020935.1_3-S - -
          Illumina Sentrix mouse-6.v1 bead array 104010440 - -
          Illumina Sentrix mouse-6.v1 bead array 102510176 - -
          Illumina Sentrix mouse-6.v1 bead array 3060451 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl020935.1_3-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102510176 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104010440 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3060451 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2753108 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2753108 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1222350 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1233178 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl020935.1_3-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline