Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00486818_m1
          See other CCT6A GE Assays ›
          Gene Symbol:
          Cct6a
          Gene Name:
          chaperonin containing Tcp1, subunit 6a (zeta)
          Gene Aliases:
          CCT-zeta, CCT-zeta-1, Cct6, Cctz-1, TCP-1-zeta
          Chromosome Location:
          Chr.5: 129787356 - 129794500 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00486818_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cct6a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009838.2 NP_033968.1 1049 80
          GenBank mRNA AK077916.1 - 1003 80
          AK134428.1 - 1002 80
          AK145522.1 - 1008 80
          AK147130.1 - 1046 80
          AK149757.1 - 1408 80
          AK150977.1 - 1003 80
          AK159354.1 - 1003 80
          AK159785.1 - 1023 80
          AK159975.1 - 1049 80
          AK160067.1 - 1003 80
          AK164312.1 - 1003 80
          AK165612.1 - 1005 80
          AK166749.1 - 1002 80
          AK166795.1 - 1032 80
          AK166870.1 - 1000 80
          AK166887.1 - 1001 80
          AK166929.1 - 1001 80
          AK167007.1 - 1031 80
          AK167076.1 - 1000 80
          AK167082.1 - 998 80
          AK167164.1 - 1008 80
          AK167180.1 - 1035 80
          AK167307.1 - 1003 80
          AK167669.1 - 1032 80
          AK167851.1 - 1004 80
          AK167994.1 - 998 80
          AK168065.1 - 998 80
          AK168940.1 - 1032 80
          AK169033.1 - 982 80
          AK180324.1 - 185 80
          AK200859.1 - 91 80
          BC018459.1 - 613 3597455 80
          BC055043.1 - 1011 80
          BC069917.1 - 1004 6394714 80
          BC094352.1 - 975 6809215 80
          Z31557.1 - 1020 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001512.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cct6a

          Panther Classification:

          Molecular Function -

          chaperonin

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          binding of sperm to zona pellucida
          toxin transport
          nucleotide binding
          protein binding
          ATP binding
          poly(A) RNA binding
          unfolded protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24090829 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24090827 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24090830 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24090828 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2125912 BC094352.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2125912 Z31557.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2125912 BC018459.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2125912 BC055043.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2125912 BC069917.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 969819 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 601377 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 250492 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 501549 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 971272 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 93538 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 5175 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116736 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2125912 BC094352.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2125912 Z31557.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2125912 BC018459.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2125912 BC055043.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2125912 BC069917.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090832 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090893 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090827 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090888 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24090882 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05743099 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05743100 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01220245 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116736 - -
          Whole Mouse Genome G4122A A_52_P363718 BC069917.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P363718 BC094352.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P363718 Z31557.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P363718 BC018459.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P363718 BC055043.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104251 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116736 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05743100 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05743099 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04394011 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04958578 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04394009 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04394013 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04864531 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04864539 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04394012 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04864529 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04864536 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04864530 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04864537 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04958575 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24090830 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24090829 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24090827 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24090828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24090896 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090865 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04115863 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090851 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090879 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090876 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090885 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090861 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090842 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090860 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090880 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090831 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04958580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090835 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04864530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090890 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090834 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090832 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090886 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090827 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090871 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090873 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090846 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090858 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090868 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04958575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04831306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24090875 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10526072 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526073 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526075 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526080 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526078 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526077 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526084 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526081 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526076 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526071 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526074 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526082 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526070 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526083 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526079 - -
          Affymetrix HT MG-430 PM Array Plate 1455988_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1423517_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10526069 - -
          Mouse Genome 430 2.0 Array 1455988_PM_a_at - -
          Mouse Genome 430 2.0 Array 1423517_PM_at - -
          Mouse Genome 430 2.0 Array 1423517_at - -
          Mouse Genome 430 2.0 Array 1455988_a_at - -
          Affymetrix Mouse Clariom S Array PSR0500013673.mm.1 TC0500001512.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500013673.mm.1 NM_009838.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500013671.mm.1 TC0500001512.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500013671.mm.1 NM_009838.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500013672.mm.1 TC0500001512.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500013672.mm.1 NM_009838.2 Refseq
          Mouse Expression Set 430 1455988_PM_a_at - -
          Mouse Expression Set 430 1423517_PM_at - -
          Mouse Expression Set 430 1423517_at - -
          Mouse Expression Set 430 1455988_a_at - -
          Murine Genome U74v2 Set 160459_r_at - -
          Murine Genome U74v2 Set 160459_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1455988_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1423517_PM_at - -
          Mouse Genome 430A 2.0 Array 1423517_at - -
          Mouse Genome 430A 2.0 Array 1455988_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1455988_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1423517_at - -
          Affymetrix HT Mouse Genome 430A Array 1455988_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1423517_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 467179 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2350368 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002350368 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2774942 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2774942 - -
          mouse-6 v1.1 (Illumina) scl27178.12.102_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 2350368 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl27178.12.102_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2350368 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2962840 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2774941 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2962840 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl27178.12.102_2-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline