Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00486893_m1
          See other CDC45 GE Assays ›
          Gene Symbol:
          Cdc45
          Gene Name:
          cell division cycle 45
          Gene Aliases:
          Cdc45l
          Chromosome Location:
          Chr.16: 18780447 - 18811972 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00486893_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdc45
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001161623.1 NP_001155095.1 6 - 7 530 89
          NM_009862.2 NP_033992.2 8 - 9 907 89
          GenBank mRNA AF081536.1 - 8 - 9 907 89
          AF081537.1 - 7 - 8 606 89
          AF081538.1 - 7 - 8 606 89
          AF098068.1 - 7 - 8 653 89
          AK147013.1 - 7 - 8 624 89
          AK167850.1 - 6 - 7 518 89
          BC006856.1 - 7 - 8 624 89
          BC028635.1 - 7 - 8 660 1262927 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600001320.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cdc45

          Panther Classification:

          Molecular Function -

          replication origin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          double-strand break repair via break-induced replication
          DNA replication
          pre-replicative complex assembly involved in nuclear cell cycle DNA replication
          DNA replication initiation
          cell cycle
          regulation of chromatin silencing at telomere
          DNA duplex unwinding
          positive regulation of G1/S transition of mitotic cell cycle
          mitotic DNA replication preinitiation complex assembly
          chromatin binding
          DNA replication origin binding
          single-stranded DNA binding
          3'-5' DNA helicase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017750 BC028635.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017750 AK147013.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017750 AK167850.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017750 BC006856.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017750 AF081538.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 354371 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 294615 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 915086 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 97423 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 440224 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 951087 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 661617 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 342992 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 634841 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 552529 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 174243 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 689379 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 865447 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 404029 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626925 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 595975 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 156240 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 333697 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 4336 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 57494 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 57495 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142948 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148090 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017750 BC028635.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017750 AK147013.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017750 AK167850.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017750 BC006856.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017750 AF081538.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P401451 BC028635.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P401451 AK147013.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P401451 AK167850.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P401451 BC006856.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P401451 AF081538.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401451 BC028635.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401451 AK147013.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401451 AK167850.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401451 BC006856.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401451 AF081538.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111699 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105982 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13006418 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13006406 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13006419 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13006405 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10438380 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438394 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438393 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438379 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438396 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438383 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438381 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438384 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438382 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438389 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438390 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438397 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438398 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438386 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438392 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438388 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438399 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438395 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438391 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438385 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438387 - -
          Affymetrix HT MG-430 PM Array Plate 1416575_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1457838_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10438378 - -
          Mouse Genome 430 2.0 Array 1416575_at - -
          Mouse Genome 430 2.0 Array 1416575_PM_at - -
          Mouse Genome 430 2.0 Array 1457838_PM_at - -
          Mouse Genome 430 2.0 Array 1457838_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12544_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12544_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1600010907.mm.1 NM_001161623.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010907.mm.1 NM_001161623.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010907.mm.1 NM_009862.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010907.mm.1 NM_009862.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010907.mm.1 TC1600001320.mm.1 MTA TC
          Mouse Expression Set 430 1416575_PM_at - -
          Mouse Expression Set 430 1416575_at - -
          Murine Genome U74v2 Set 102403_PM_at - -
          Murine Genome U74v2 Set 102403_at - -
          Murine Genome U74v2 Set 171199_PM_r_at - -
          Murine Genome U74v2 Set 171199_r_at - -
          Mouse Genome 430A 2.0 Array 1416575_PM_at - -
          Mouse Genome 430A 2.0 Array 1416575_at - -
          Affymetrix HT Mouse Genome 430A Array 1416575_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416575_at - -
          Affymetrix HT Mouse Genome 430B Array 1457838_at - -
          Affymetrix HT Mouse Genome 430B Array 1457838_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 906073 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 70537 - -
          Illumina mouse-6 v1.0 expression beadchip 70537 - -
          Illumina mouse-6 v1.0 expression beadchip 3130114 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000070537 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003130114 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1233450 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2622551 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2622551 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1233450 - -
          mouse-6 v1.1 (Illumina) scl0001902.1_2-S - -
          mouse-6 v1.1 (Illumina) scl48677.19.2_5-S - -
          Illumina Sentrix mouse-6.v1 bead array 3130114 - -
          Illumina Sentrix mouse-6.v1 bead array 70537 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001902.1_2-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl48677.19.2_5-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 70537 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3130114 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2622551 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2919433 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2622551 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2919433 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline