Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00487041_m1
          See other COL10A1 GE Assays ›
          Gene Symbol:
          Col10a1
          Gene Name:
          collagen, type X, alpha 1
          Gene Aliases:
          Col10, Col10a-1
          Chromosome Location:
          Chr.10: 34389981 - 34397085 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00487041_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Col10a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009925.4 NP_034055.1 2 - 3 223 77
          GenBank mRNA BY141606.1 - 2 - 3 224 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000000300.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Col10a1

          Panther Classification:

          Molecular Function -

          extracellular matrix structural protein

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2089520 CJ326431.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2089520 NM_009925.4 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P144492 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2089520 CJ326431.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2089520 NM_009925.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P289032 X63013.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P289032 NM_009925.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056726 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056728 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056731 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056724 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056729 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27056738 - -
          Whole Mouse Genome G4122A A_51_P289032 X63013.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P289032 NM_009925.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131447 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08821506 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08821523 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08821510 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08821497 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08821519 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08821515 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08821498 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08821522 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08821516 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08821527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27056754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08456650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08456637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056744 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08283556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08462241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08290628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08462253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056814 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08422870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08290618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056722 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08821503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08422869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08462244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08505902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27056742 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10362496 - -
          Affymetrix Mouse Gene 1.0 ST Array 10362497 - -
          Affymetrix Mouse Gene 1.0 ST Array 10362498 - -
          Affymetrix HT MG-430 PM Array Plate 1422253_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10362495 - -
          Mouse Genome 430 2.0 Array 1422253_PM_at - -
          Mouse Genome 430 2.0 Array 1422253_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12813_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12813_at - -
          Affymetrix Mouse Clariom S Array PSR1000001999.mm.1 NM_009925.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001999.mm.1 NM_009925.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001999.mm.1 TC1000000300.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000001998.mm.1 NM_009925.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001998.mm.1 NM_009925.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001998.mm.1 TC1000000300.mm.1 MTA TC
          Mouse Expression Set 430 1422253_at - -
          Mouse Expression Set 430 1422253_PM_at - -
          Murine Genome U74v2 Set 92921_at - -
          Murine Genome U74v2 Set 92921_PM_at - -
          Mouse Genome 430A 2.0 Array 1422253_at - -
          Mouse Genome 430A 2.0 Array 1422253_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422253_at - -
          Affymetrix HT Mouse Genome 430A Array 1422253_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 378029 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 840059 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000840059 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2675386 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2675386 - -
          mouse-6 v1.1 (Illumina) scl39030.3_572-S - -
          Illumina Sentrix mouse-6.v1 bead array 840059 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl39030.3_572-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 840059 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2675388 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2675388 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline