Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00487230_g1
          See other CYP21A1 GE Assays ›
          Gene Symbol:
          Cyp21a1
          Gene Name:
          cytochrome P450, family 21, subfamily a, polypeptide 1
          Gene Aliases:
          21-OH, 21OH, 21OHA, 21OHB, CYP21OH-A, Cyp21, Cyp21-ps1, Cyp21B, Cyp21a2-ps, Cyp21a2ps, Oh21-1, Oh21-2
          Chromosome Location:
          Chr.17: 34801348 - 34804426 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00487230_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp21a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009995.2 NP_034125.2 1 - 2 233 76
          GenBank mRNA AK146565.1 - 1 - 2 370 76
          BC127167.1 - 1 - 2 206 40105690 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001927.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection P450 Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyp21a1

          Gene Ontology Categories:

          Function(s) Process(es)

          steroid biosynthetic process
          C21-steroid hormone biosynthetic process
          mineralocorticoid biosynthetic process
          progesterone metabolic process
          oxidation-reduction process
          monooxygenase activity
          steroid 21-monooxygenase activity
          steroid binding
          iron ion binding
          lipid binding
          oxidoreductase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
          heme binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2171942 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2140107 NM_009995.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2140107 AK146565.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2140107 BQ033552.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 750383 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 836415 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102010 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131724 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2171942 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2140107 NM_009995.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2140107 AK146565.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2140107 BQ033552.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P594116 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155866 - -
          Whole Mouse Genome G4122A A_52_P594116 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116421 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13310763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13196972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155877 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13474774 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13081206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13474777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13474738 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155798 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13081209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13474801 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10450279 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450275 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450273 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450277 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450274 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450278 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450276 - -
          Affymetrix HT MG-430 PM Array Plate 1455691_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1422333_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10450272 - -
          Mouse Genome 430 2.0 Array 1422333_at - -
          Mouse Genome 430 2.0 Array 1455691_PM_at - -
          Mouse Genome 430 2.0 Array 1455691_at - -
          Mouse Genome 430 2.0 Array 1422333_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13079_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13079_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700018120.mm.1 NM_009995.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018120.mm.1 NM_009995.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018120.mm.1 TC1700001927.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018118.mm.1 NM_009995.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018118.mm.1 NM_009995.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018118.mm.1 TC1700001927.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018121.mm.1 NM_009995.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018121.mm.1 NM_009995.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018121.mm.1 TC1700001927.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018123.mm.1 NM_009995.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018123.mm.1 NM_009995.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018123.mm.1 TC1700001927.mm.1 MTA TC
          Mouse Expression Set 430 1455691_at - -
          Mouse Expression Set 430 1455691_PM_at - -
          Mouse Expression Set 430 1422333_PM_at - -
          Mouse Expression Set 430 1422333_at - -
          Murine Genome U74v2 Set 93225_s_at - -
          Murine Genome U74v2 Set 93225_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1455691_at - -
          Mouse Genome 430A 2.0 Array 1422333_at - -
          Mouse Genome 430A 2.0 Array 1422333_PM_at - -
          Mouse Genome 430A 2.0 Array 1455691_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1455691_at - -
          Affymetrix HT Mouse Genome 430A Array 1455691_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422333_at - -
          Affymetrix HT Mouse Genome 430A Array 1422333_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 534285 - -
          ABI Mouse Genome Survey Microarray 624698 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4060427 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004060427 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2673575 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2673575 - -
          mouse-6 v1.1 (Illumina) scl50014.7.1_3-S - -
          Illumina Sentrix mouse-6.v1 bead array 4060427 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50014.7.1_3-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4060427 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2673575 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2879008 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2879008 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2673575 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline