Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00487425_m1
          See other FOS GE Assays ›
          Gene Symbol:
          Fos
          Gene Name:
          FBJ osteosarcoma oncogene
          Gene Aliases:
          D12Rfj1, c-fos, cFos
          Chromosome Location:
          Chr.12: 85473901 - 85477269 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00487425_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (10)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fos
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010234.2 NP_034364.1 1 - 2 279 59
          GenBank mRNA AK154418.1 - 1 - 2 291 59
          BC029814.1 - 1 - 2 279 2582234 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200000833.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fos

          Panther Classification:

          Molecular Function -

          basic leucine zipper transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          transforming growth factor beta receptor signaling pathway
          nervous system development
          cellular response to extracellular stimulus
          cellular response to reactive oxygen species
          skeletal muscle cell differentiation
          response to muscle stretch
          response to drug
          positive regulation of osteoclast differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          SMAD protein signal transduction
          cellular response to calcium ion
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          chromatin binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          transcription factor binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          protein heterodimerization activity
          R-SMAD binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200006061.mm.1 NM_010234.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200006061.mm.1 NM_010234.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200006061.mm.1 TC1200000833.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200006057.mm.1 NM_010234.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200006057.mm.1 NM_010234.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200006057.mm.1 TC1200000833.mm.1 MTA TC

          Back To Top
           

          Citations (10)

          Alterations in gene regulation following inhibition of the striatum-enriched phosphodiesterase, PDE10A.
          Authors
          Strick CA, James LC, Fox CB, Seeger TF, Menniti FS, Schmidt CJ
          Journal
          Neuropharmacology - Page No 444 (Mon Feb 01 00:00:00 EST 2010)
          Volume (Year)
          58 (2010)
          Product Usage
          -
          Comparative analysis of gene expression changes mediated by individual constituents of hemozoin.
          Authors
          Schrimpe AC, Wright DW
          Journal
          Chem Res Toxicol - Page No 433 (Mon Mar 16 00:00:00 EDT 2009)
          Volume (Year)
          22 (2009)
          Product Usage
          -
          Leptin regulates chondrogenic differentiation in ATDC5 cell-line through JAK/STAT and MAPK pathways.
          Authors
          Ben-Eliezer M, Phillip M, Gat-Yablonski G
          Journal
          Endocrine - Page No 235 (Mon Oct 01 00:00:00 EDT 2007)
          Volume (Year)
          32 (2007)
          Product Usage
          -
          Tenascin-C induction by cyclic strain requires integrin-linked kinase.
          Authors
          Maier S, Lutz R, Gelman L, Sarasa-Renedo A, Schenk S, Grashoff C, Chiquet M
          Journal
          Biochim Biophys Acta - Page No 1150 (Sun Jun 01 00:00:00 EDT 2008)
          Volume (Year)
          1783 (2008)
          Product Usage
          -
          ErbB activity links the glucagon-like peptide-2 receptor to refeeding-induced adaptation in the murine small bowel.
          Authors
          Bahrami J, Yusta B, Drucker DJ
          Journal
          Gastroenterology - Page No 2447 (Tue Jun 01 00:00:00 EDT 2010)
          Volume (Year)
          138 (2010)
          Product Usage
          -
          Tough beginnings: alterations in the transcriptome of cloned embryos during the first two cell cycles.
          Authors
          Vassena R, Han Z, Gao S, Baldwin DA, Schultz RM, Latham KE
          Journal
          Dev Biol - Page No 75 (Sun Apr 01 00:00:00 EDT 2007)
          Volume (Year)
          304 (2007)
          Product Usage
          -
          Long-term depression activates transcription of immediate early transcription factor genes: involvement of serum response factor/Elk-1.
          Authors
          Lindecke A, Korte M, Zagrebelsky M, Horejschi V, Elvers M, Widera D, Prullage M, Pfeiffer J, Kaltschmidt B, Kaltschmidt C
          Journal
          Eur J Neurosci - Page No 555 (Sat Jul 01 00:00:00 EDT 2006)
          Volume (Year)
          24 (2006)
          Product Usage
          -
          BHLHB2 controls Bdnf promoter 4 activity and neuronal excitability.
          Authors
          Jiang X, Tian F, Du Y, Copeland NG, Jenkins NA, Tessarollo L, Wu X, Pan H, Hu XZ, Xu K, Kenney H, Egan SE, Turley H, Harris AL, Marini AM, Lipsky RH
          Journal
          J Neurosci - Page No 1118 (Wed Jan 30 00:00:00 EST 2008)
          Volume (Year)
          28 (2008)
          Product Usage
          -
          Control of the establishment of aversive memory by calcineurin and Zif268.
          Authors
          Baumgartel K, Genoux D, Welzl H, Tweedie-Cullen RY, Koshibu K, Livingstone-Zatchej M, Mamie C, Mansuy IM
          Journal
          Nat Neurosci - Page No 572 (Thu May 01 00:00:00 EDT 2008)
          Volume (Year)
          11 (2008)
          Product Usage
          -
          The influence of diesel exhaust on polycyclic aromatic hydrocarbon-induced DNA damage, gene expression, and tumor initiation in Sencar mice in vivo.
          Authors
          Courter LA, Luch A, Musafia-Jeknic T, Arlt VM, Fischer K, Bildfell R, Pereira C, Phillips DH, Poirier MC, Baird WM
          Journal
          Cancer Lett - Page No 135 (Sat Jun 28 00:00:00 EDT 2008)
          Volume (Year)
          265 (2008)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline