Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00487473_m1
          See other GAMT GE Assays ›
          Gene Symbol:
          Gamt
          Gene Name:
          guanidinoacetate methyltransferase
          Gene Aliases:
          AA571402, Spintz1
          Chromosome Location:
          Chr.10: 80258151 - 80261024 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00487473_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gamt
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010255.3 NP_034385.1 1 - 2 249 105
          GenBank mRNA AA881765.1 - 1 - 2 267 106
          AF015887.1 - 1 - 2 182 105
          AK140688.1 - 1 - 2 250 105
          AK158087.1 - 1 - 2 192 105
          BC049233.1 - 1 - 2 235 4195030 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002493.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gamt

          Panther Classification:

          Molecular Function -

          methyltransferase transferase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          creatine biosynthetic process
          spermatogenesis
          organ morphogenesis
          methylation
          regulation of multicellular organism growth
          S-adenosylhomocysteine metabolic process
          S-adenosylmethionine metabolic process
          methyltransferase activity
          S-adenosylmethionine-dependent methyltransferase activity
          transferase activity
          guanidinoacetate N-methyltransferase activity
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27282058 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27282041 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27282039 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27282040 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1973259 AK158087.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1973259 AK217844.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1973259 BC049233.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1973259 BE992264.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1973259 NM_010255.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1991505 BE992264.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1991505 NM_010255.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 492533 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 220112 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105125 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1991505 BE992264.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1991505 NM_010255.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1973259 AK158087.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1973259 AK217844.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1973259 BC049233.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1973259 BE992264.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1973259 NM_010255.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P484869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282032 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105125 - -
          Whole Mouse Genome G4122A A_51_P484869 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130270 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105125 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08361804 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08564651 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08361798 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08361802 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08361811 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08222367 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08361809 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08879565 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08564652 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08361797 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27282039 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27282040 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27282041 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27282058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27281963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08222365 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08361803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08902215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08222367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08356725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08356712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282045 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10370772 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370768 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370767 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370773 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370771 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370769 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370775 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370774 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370770 - -
          Affymetrix HT MG-430 PM Array Plate 1422558_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10370766 - -
          Mouse Genome 430 2.0 Array 1422558_PM_at - -
          Mouse Genome 430 2.0 Array 1422558_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14431_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14431_at - -
          Affymetrix Mouse Clariom S Array PSR1000018399.mm.1 NM_010255.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018399.mm.1 TC1000002493.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018400.mm.1 NM_010255.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018400.mm.1 TC1000002493.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018401.mm.1 NM_010255.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018401.mm.1 TC1000002493.mm.1 MTA TC
          Mouse Expression Set 430 1422558_at - -
          Mouse Expression Set 430 1422558_PM_at - -
          Murine Genome U74v2 Set 161644_PM_f_at - -
          Murine Genome U74v2 Set 161644_f_at - -
          Murine Genome U74v2 Set 101408_at - -
          Murine Genome U74v2 Set 101408_PM_at - -
          Mouse Genome 430A 2.0 Array 1422558_at - -
          Mouse Genome 430A 2.0 Array 1422558_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422558_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422558_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 930389 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6980301 - -
          Illumina mouse-6 v1.0 expression beadchip 6980301 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006980301 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2757391 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2757391 - -
          mouse-6 v1.1 (Illumina) scl37740.6.1_17-S - -
          Illumina Sentrix mouse-6.v1 bead array 6980301 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37740.6.1_17-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6980301 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2815506 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2815506 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl37740.6.1_17-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline