Search
Search
| Gene Symbol: Mmp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010810.4 | NP_034940.2 | 1 - 2 | 132 | 128 | |
| GenBank mRNA | AK144515.1 | - | 1 - 2 | 150 | 128 | |
| BC119057.1 | - | 1 - 2 | 132 | 8733806 | 128 | |
| BC120655.1 | - | 1 - 2 | 132 | 40129578 | 128 | |
| L36244.1 | - | 1 - 2 | 157 | 128 | ||
Set Membership: |
|