Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00487796_m1
          See other MX1 GE Assays ›
          Gene Symbol:
          Mx1
          Gene Name:
          MX dynamin-like GTPase 1
          Gene Aliases:
          AI893580, Mx, Mx-1
          Chromosome Location:
          Chr.16: 97447035 - 97462906 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00487796_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mx1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010846.1 NP_034976.1 1619 121
          NR_003520.1 7 - 8 1100 121
          GenBank mRNA AK076430.1 - 12 - 13 1371 121
          AK150023.1 - 7 - 8 1099 121
          AK150236.1 - 7 - 8 1099 121
          BC011113.1 - 1604 4011821 121
          CT010406.1 - 1406 121
          EU638327.1 - 3 - 4 424 121
          EU638328.1 - 6 - 7 917 121
          M12279.1 - 1619 121
          M21038.1 - 1691 121
          M21039.1 - 9 - 10 1195 121

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mx1

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          response to virus
          innate immune response
          defense response to virus
          nucleotide binding
          GTPase activity
          protein binding
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2118441 M21039.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2118441 AK150023.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2118441 AK150236.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2118441 NR_003520.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2118441 AK076430.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 934989 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 194199 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 679588 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 842246 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 352828 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 964409 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 373794 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 206180 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 34443 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 60015 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 151658 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108908 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128409 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2118441 M21039.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2118441 AK150023.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2118441 AK150236.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2118441 NR_003520.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2118441 AK076430.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999095 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999471 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999100 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999475 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999101 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999077 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999482 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30999496 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P128409 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P108908 - -
          Whole Mouse Genome G4122A A_52_P614259 M21039.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P614259 M12279.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P614259 M21038.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P614259 CT010406.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P614259 EU638327.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P446431 M21039.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P446431 M12279.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P446431 M21038.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P446431 BC011113.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P446431 AK150023.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107451 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131542 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P128409 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P108908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30998970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30998961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30998977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30998976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30998972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30998998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30998950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30998974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30998982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30998952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30998966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30998965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30998993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30998953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30998957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30998959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30998973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30998946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30998989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30998983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30998988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30998986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30998995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30999012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12655191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30999058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30999042 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12599035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30999101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30999040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12494475 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30999107 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30999055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12696299 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30999045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30998952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30999039 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30999075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30999052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12655227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30999070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12655186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30999077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12645424 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30999047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12655198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12833520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30999090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30999100 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30999088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30999104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30999092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30999087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30999032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30999095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30999080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30999097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12800464 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30999050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30999086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30998948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30999089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13025136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12655223 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12655185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30999108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30998950 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10441243 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441239 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441236 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441234 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441242 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441240 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441235 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441237 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441241 - -
          Affymetrix HT MG-430 PM Array Plate 1451905_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10441233 - -
          Mouse Genome 430 2.0 Array 1451905_a_at - -
          Mouse Genome 430 2.0 Array 1451905_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17857_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17857_at - -
          Mouse Expression Set 430 1451905_PM_a_at - -
          Mouse Expression Set 430 1451905_a_at - -
          Murine Genome U74v2 Set 98417_PM_at - -
          Murine Genome U74v2 Set 98417_at - -
          Mouse Genome 430A 2.0 Array 1451905_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1451905_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1451905_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1451905_a_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2690301 - -
          Illumina mouse-6 v1.0 expression beadchip 6840364 - -
          Illumina mouse-6 v1.0 expression beadchip 2690301 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002690301 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006840364 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2771610 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2707870 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2707870 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2771610 - -
          mouse-6 v1.1 (Illumina) scl017857.1_53-S - -
          mouse-6 v1.1 (Illumina) scl48151.12.1_9-S - -
          Illumina Sentrix mouse-6.v1 bead array 6840364 - -
          Illumina Sentrix mouse-6.v1 bead array 2690301 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl017857.1_53-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl48151.12.1_9-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2690301 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6840364 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2707870 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2707870 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline