Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00488055_m1
          See other RPGR GE Assays ›
          Gene Symbol:
          Rpgr
          Gene Name:
          retinitis pigmentosa GTPase regulator
          Gene Aliases:
          Rp3h
          Chromosome Location:
          Chr.X: 10158216 - 10216795 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00488055_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpgr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001177950.1 NP_001171421.1 3 - 4 465 66
          NM_001177951.1 NP_001171422.1 3 - 4 465 66
          NM_001177952.1 NP_001171423.1 3 - 4 459 66
          NM_001177953.1 NP_001171424.1 3 - 4 465 66
          NM_001177954.1 NP_001171425.1 3 - 4 465 66
          NM_011285.2 NP_035415.1 3 - 4 465 66
          GenBank mRNA AF044677.1 - 3 - 4 383 66
          AJ238396.1 - 3 - 4 377 66
          AK017192.1 - 3 - 4 397 66
          AK029864.1 - 3 - 4 591 66
          AK038652.1 - 3 - 4 429 66
          AK081631.1 - 3 - 4 404 66
          AK136298.1 - 3 - 4 396 66
          AK138977.1 - 3 - 4 460 66
          BC012661.1 - 3 - 4 404 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00003418.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpgr

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          visual perception
          cell projection organization
          intraciliary transport
          cilium assembly
          eye photoreceptor cell development
          response to stimulus
          retina morphogenesis in camera-type eye
          cellular response to light stimulus
          guanyl-nucleotide exchange factor activity
          protein binding
          poly(A) RNA binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0X00011796.mm.1 NM_001177950.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011796.mm.1 NM_001177950.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011796.mm.1 NM_001177951.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011796.mm.1 NM_001177951.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011796.mm.1 NM_001177952.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011796.mm.1 NM_001177952.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011796.mm.1 NM_001177953.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011796.mm.1 NM_001177953.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011796.mm.1 NM_001177954.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011796.mm.1 NM_001177954.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011796.mm.1 NM_011285.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011796.mm.1 NM_011285.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011796.mm.1 TC0X00003418.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 608106 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline