Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00488099_g1
          See other APCS GE Assays ›
          Gene Symbol:
          Apcs
          Gene Name:
          serum amyloid P-component
          Gene Aliases:
          Sap
          Chromosome Location:
          Chr.1: 172893961 - 172895054 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00488099_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Apcs
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011318.2 NP_035448.2 1 - 2 232 114
          GenBank mRNA AK144248.1 - 1 - 2 119 114
          AK165699.1 - 1 - 2 119 114
          BC061125.1 - 1 - 2 129 30308397 114
          M23552.1 - 1 - 2 208 114
          X14079.1 - 1 - 2 208 114
          Y00426.1 - 1 - 2 221 114

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100003580.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Mouse Alzheimer's Disease 1700 Inventoried Mouse_Alzheimer Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Apcs

          Gene Ontology Categories:

          Function(s) Process(es)

          protein complex assembly
          negative regulation by host of viral exo-alpha-sialidase activity
          negative regulation by host of viral glycoprotein metabolic process
          innate immune response
          negative regulation of monocyte differentiation
          negative regulation of viral entry into host cell
          negative regulation of viral process
          negative regulation of exo-alpha-sialidase activity
          negative regulation of glycoprotein metabolic process
          complement component C1q binding
          calcium ion binding
          carbohydrate binding
          virion binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P354165 M23552.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P354165 X14079.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P354165 Y00426.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P354165 M23552.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P354165 X14079.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P354165 Y00426.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 428441 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P130498 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P354165 M23552.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P354165 X14079.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P354165 Y00426.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P354165 M23552.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P354165 X14079.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P354165 Y00426.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P354165 M23552.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P354165 X14079.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P354165 Y00426.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P354165 M23552.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P354165 X14079.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P354165 Y00426.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835475 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835474 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835472 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835470 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835480 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835477 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835466 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835494 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P130498 - -
          Whole Mouse Genome G4122A A_51_P354165 M23552.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P354165 X14079.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P354165 Y00426.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P354165 M23552.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P354165 X14079.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P354165 Y00426.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104939 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129074 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P130498 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00801132 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00801115 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00801131 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00801118 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00801125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835464 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835513 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835492 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00057412 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835499 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00409229 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835542 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835479 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835487 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835477 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00909489 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835475 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835544 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835489 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00409227 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835495 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835459 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835501 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835486 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835481 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835453 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835480 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835462 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835498 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835509 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10360330 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360331 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360329 - -
          Affymetrix HT MG-430 PM Array Plate 1419059_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10360328 - -
          Mouse Genome 430 2.0 Array 1419059_PM_at - -
          Mouse Genome 430 2.0 Array 1419059_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20219_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20219_PM_at - -
          Affymetrix Murine Genome 6500A Array Msa11630_at - -
          Affymetrix Mouse Clariom S Array PSR0100029388.mm.1 NM_011318.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029388.mm.1 NM_011318.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029388.mm.1 TC0100003580.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100029389.mm.1 NM_011318.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029389.mm.1 NM_011318.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029389.mm.1 TC0100003580.mm.1 MTA TC
          Mouse Expression Set 430 1419059_at - -
          Mouse Expression Set 430 1419059_PM_at - -
          Murine Genome U74v2 Set 104072_PM_at - -
          Murine Genome U74v2 Set 104072_at - -
          Mouse Genome 430A 2.0 Array 1419059_at - -
          Mouse Genome 430A 2.0 Array 1419059_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419059_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419059_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 926486 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2060500 - -
          Illumina mouse-6 v1.0 expression beadchip 2060500 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002060500 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2593329 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2593329 - -
          mouse-6 v1.1 (Illumina) scl15913.2.1_30-S - -
          Illumina Sentrix mouse-6.v1 bead array 2060500 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl15913.2.1_30-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2060500 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2896170 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2896170 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline