Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00488139_m1
          See other SERF1 GE Assays ›
          Gene Symbol:
          Serf1
          Gene Name:
          small EDRK-rich factor 1
          Gene Aliases:
          4F5, Msmac1, m4F5
          Chromosome Location:
          Chr.13: 100108019 - 100114233 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00488139_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011353.2 NP_035483.1 2 - 3 319 66
          GenBank mRNA AF073517.1 - 2 - 3 290 66
          AK005773.1 - 2 - 3 234 66
          BC028510.1 - 2 - 3 272 1510984 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300001135.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Serf1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29267217 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29267241 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29267214 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29267218 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29267215 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29267238 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29267216 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29267242 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254986 AK005773.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254986 BC028510.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254986 NM_011353.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254986 AF073517.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254986 AK005773.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254986 BC028510.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254986 NM_011353.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254986 AF073517.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 782977 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 598901 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 151260 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 228264 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 69466 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153346 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254986 AK005773.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254986 BC028510.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254986 NM_011353.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254986 AF073517.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254986 AK005773.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254986 BC028510.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254986 NM_011353.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254986 AF073517.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29267252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29267231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29267211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29267255 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29267213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29267227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29267218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29267215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29267229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29267235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29267254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29267216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29267244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29267221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29267242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29267238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29267210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29267249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29267223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29267217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29267241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29267237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29267214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29267246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29267247 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29267224 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153346 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07971622 - -
          Whole Mouse Genome G4122A A_51_P254986 AK005773.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254986 BC028510.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254986 NM_011353.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254986 AF073517.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254986 AK005773.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254986 BC028510.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254986 NM_011353.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254986 AF073517.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109361 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153346 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10631171 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29267214 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29267216 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29267218 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29267215 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29267217 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29267241 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29267238 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29267242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29267266 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29267247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29267237 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29267233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10678496 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29267257 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29267254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10965905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29267260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29267252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29267249 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29267270 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29267243 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10652596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29267250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29267238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29267276 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29267229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29267230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10786832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29267246 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10631171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29267262 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29267264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10846681 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29267235 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10611832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29267272 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10411594 - -
          Affymetrix Mouse Gene 1.0 ST Array 10406880 - -
          Affymetrix Mouse Gene 1.0 ST Array 10406879 - -
          Affymetrix Mouse Gene 1.0 ST Array 10406878 - -
          Affymetrix HT MG-430 PM Array Plate 1449509_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1454908_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1454907_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1434291_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10406877 - -
          Affymetrix Mouse Gene 1.1 ST Array 10411593 - -
          Mouse Genome 430 2.0 Array 1454908_x_at - -
          Mouse Genome 430 2.0 Array 1454907_PM_at - -
          Mouse Genome 430 2.0 Array 1449509_PM_at - -
          Mouse Genome 430 2.0 Array 1454908_PM_x_at - -
          Mouse Genome 430 2.0 Array 1452502_PM_at - -
          Mouse Genome 430 2.0 Array 1434291_a_at - -
          Mouse Genome 430 2.0 Array 1434291_PM_a_at - -
          Mouse Genome 430 2.0 Array 1452502_at - -
          Mouse Genome 430 2.0 Array 1449509_at - -
          Mouse Genome 430 2.0 Array 1454907_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20365_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20365_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300007199.mm.1 NM_011353.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300007199.mm.1 NM_011353.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300007199.mm.1 TC1300001135.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300007200.mm.1 NM_011353.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300007200.mm.1 NM_011353.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300007200.mm.1 TC1300001135.mm.1 MTA TC
          Mouse Expression Set 430 1449509_at - -
          Mouse Expression Set 430 1454907_at - -
          Mouse Expression Set 430 1434291_a_at - -
          Mouse Expression Set 430 1454907_PM_at - -
          Mouse Expression Set 430 1454908_PM_x_at - -
          Mouse Expression Set 430 1434291_PM_a_at - -
          Mouse Expression Set 430 1454908_x_at - -
          Mouse Expression Set 430 1449509_PM_at - -
          Murine Genome U74v2 Set 103094_PM_at - -
          Murine Genome U74v2 Set 103094_at - -
          Mouse Genome 430A 2.0 Array 1454907_at - -
          Mouse Genome 430A 2.0 Array 1449509_at - -
          Mouse Genome 430A 2.0 Array 1454907_PM_at - -
          Mouse Genome 430A 2.0 Array 1454908_x_at - -
          Mouse Genome 430A 2.0 Array 1434291_a_at - -
          Mouse Genome 430A 2.0 Array 1449509_PM_at - -
          Mouse Genome 430A 2.0 Array 1454908_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1434291_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1434291_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1449509_at - -
          Affymetrix HT Mouse Genome 430A Array 1449509_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1454908_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1454907_at - -
          Affymetrix HT Mouse Genome 430A Array 1454907_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1434291_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1454908_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 933097 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2260731 - -
          Illumina mouse-6 v1.0 expression beadchip 6980075 - -
          Illumina mouse-6 v1.0 expression beadchip 2260731 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006980075 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002260731 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2631518 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2747109 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2631518 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2747109 - -
          mouse-6 v1.1 (Illumina) scl44462.5.1_15-S - -
          mouse-6 v1.1 (Illumina) scl0020365.1_201-S - -
          Illumina Sentrix mouse-6.v1 bead array 2260731 - -
          Illumina Sentrix mouse-6.v1 bead array 6980075 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl44462.5.1_15-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0020365.1_201-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2260731 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6980075 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2631518 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2747109 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2747109 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2631518 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0020365.1_201-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline