Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc12a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011390.2 | NP_035520.1 | 4 - 5 | 558 | 58 | |
GenBank mRNA | AF087436.1 | - | 4 - 5 | 560 | 58 | |
AK052910.1 | - | 4 - 5 | 609 | 58 | ||
AK131129.1 | - | 4 - 5 | 512 | 58 | ||
AK143535.1 | - | 4 - 5 | 552 | 58 | ||
AK149750.1 | - | 4 - 5 | 553 | 58 | ||
AK166215.1 | - | 4 - 5 | 552 | 58 | ||
AK169950.1 | - | 4 - 5 | 548 | 58 | ||
AK171498.1 | - | 4 - 5 | 595 | 58 | ||
BC051558.1 | - | 4 - 5 | 547 | 58 | ||
BC053470.1 | - | 4 - 5 | 1184 | 58 | ||
BC059242.1 | - | 4 - 5 | 550 | 58 | ||
BC060133.1 | - | 4 - 5 | 550 | 58 | ||
BC079677.1 | - | 4 - 5 | 516 | 58 | ||
BC141107.1 | - | 4 - 5 | 518 | 9055719 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |