Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00488307_m1
          See other SLFN2 GE Assays ›
          Gene Symbol:
          Slfn2
          Gene Name:
          schlafen 2
          Gene Aliases:
          Shlf2
          Chromosome Location:
          Chr.11: 83065112 - 83070678 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00488307_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slfn2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011408.1 NP_035538.1 1 - 2 229 65
          GenBank mRNA AF099973.1 - 1 - 2 229 65
          BC138749.1 - 1 - 2 184 8861771 65
          BC138750.1 - 1 - 2 184 8861772 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001238.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slfn2

          Gene Ontology Categories:

          Function(s)

          negative regulation of cell proliferation

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P423578 AF099973.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P423578 BC138749.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P423578 BC138750.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P423578 NM_011408.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P423578 AF099973.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P423578 BC138749.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P423578 BC138750.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P423578 NM_011408.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 387642 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 637184 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 35359 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 153733 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 6907 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155677 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P423578 AF099973.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P423578 BC138749.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P423578 BC138750.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P423578 NM_011408.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P423578 AF099973.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P423578 BC138749.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P423578 BC138750.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P423578 NM_011408.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969832 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969801 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969820 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969800 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969805 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969809 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969795 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969814 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969807 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969823 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969837 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02379094 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155677 - -
          Whole Mouse Genome G4122A A_51_P423578 AF099973.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P423578 BC138749.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P423578 BC138750.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P423578 NM_011408.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P423578 AF099973.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P423578 BC138749.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P423578 BC138750.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P423578 NM_011408.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106286 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111565 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155677 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09295345 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09295346 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09128637 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09295344 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09128636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09549576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09275944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09691922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09623552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09696111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09345002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09295340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969814 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09275956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09219611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09128639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09245153 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10379632 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379631 - -
          Affymetrix HT MG-430 PM Array Plate 1450165_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10379630 - -
          Mouse Genome 430 2.0 Array 1450165_at - -
          Mouse Genome 430 2.0 Array 1450165_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20556_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20556_at - -
          Affymetrix Mouse Clariom S Array PSR1100011625.mm.1 NM_011408.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011625.mm.1 NM_011408.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011625.mm.1 TC1100001238.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100011626.mm.1 NM_011408.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011626.mm.1 NM_011408.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011626.mm.1 TC1100001238.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100011624.mm.1 NM_011408.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011624.mm.1 NM_011408.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011624.mm.1 TC1100001238.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100011628.mm.1 NM_011408.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011628.mm.1 NM_011408.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011628.mm.1 TC1100001238.mm.1 MTA TC
          Mouse Expression Set 430 1450165_at - -
          Mouse Expression Set 430 1450165_PM_at - -
          Murine Genome U74v2 Set 92472_f_at - -
          Murine Genome U74v2 Set 92471_PM_i_at - -
          Murine Genome U74v2 Set 92471_i_at - -
          Murine Genome U74v2 Set 92472_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1450165_PM_at - -
          Mouse Genome 430A 2.0 Array 1450165_at - -
          Affymetrix HT Mouse Genome 430A Array 1450165_at - -
          Affymetrix HT Mouse Genome 430A Array 1450165_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 546299 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3120114 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003120114 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1219074 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1219074 - -
          mouse-6 v1.1 (Illumina) scl41148.2_393-S - -
          Illumina Sentrix mouse-6.v1 bead array 3120114 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41148.2_393-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3120114 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1219074 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1219074 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl41148.2_393-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0