Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00488318_m1
          See other SMPD1 GE Assays ›
          Gene Symbol:
          Smpd1
          Gene Name:
          sphingomyelin phosphodiesterase 1, acid lysosomal
          Gene Aliases:
          A-SMase, ASM, Zn-SMase, aSMase
          Chromosome Location:
          Chr.7: 105554360 - 105558389 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00488318_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smpd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011421.2 NP_035551.1 2 - 3 1257 129
          GenBank mRNA AK145534.1 - 2 - 3 1226 129
          AK145702.1 - 2 - 3 1258 129
          AK149579.1 - 2 - 3 1226 129
          AK164167.1 - 2 - 3 1226 129
          AK164717.1 - 2 - 3 1181 129
          AK167802.1 - 2 - 3 1180 129
          BC011304.1 - 2 - 3 1192 3964394 129
          BC034515.1 - 2 - 3 1182 4482098 129
          Z14252.1 - 2 - 3 1203 129

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001554.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Smpd1

          Panther Classification:

          Molecular Function -

          hydrolase phosphodiesterase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          sphingomyelin catabolic process
          metabolic process
          termination of signal transduction
          positive regulation of protein dephosphorylation
          response to cocaine
          positive regulation of apoptotic process
          negative regulation of MAP kinase activity
          ceramide biosynthetic process
          sphingomyelin phosphodiesterase activity
          hydrolase activity
          hydrolase activity, acting on glycosyl bonds

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P429209 Z14252.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P429209 BC011304.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P429209 BC034515.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P429209 BP760033.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P429209 AK205584.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P429209 Z14252.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P429209 BC011304.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P429209 BC034515.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P429209 BP760033.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P429209 AK205584.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 370559 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 73708 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 187312 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 126429 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P180516 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P429209 Z14252.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P429209 BC011304.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P429209 BC034515.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P429209 BP760033.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P429209 AK205584.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P429209 Z14252.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P429209 BC011304.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P429209 BC034515.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P429209 BP760033.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P429209 AK205584.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P429209 Z14252.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P429209 BC011304.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P429209 BC034515.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P429209 BP760033.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P429209 AK205584.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P429209 Z14252.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P429209 BC011304.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P429209 BC034515.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P429209 BP760033.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P429209 AK205584.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392904 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392922 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P180516 - -
          Whole Mouse Genome G4122A A_51_P429209 Z14252.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P429209 BC011304.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P429209 BC034515.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P429209 BP760033.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P429209 AK205584.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P429209 Z14252.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P429209 BC011304.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P429209 BC034515.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P429209 BP760033.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P429209 AK205584.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140696 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P180516 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05950400 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05950428 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06236174 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05950422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06661442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06626196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06661443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06374644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06236174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06231415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05950401 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06374641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05950419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06626194 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555948 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555952 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555947 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555951 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555950 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555949 - -
          Affymetrix HT MG-430 PM Array Plate 1448621_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1447874_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10555946 - -
          Mouse Genome 430 2.0 Array 1447874_PM_x_at - -
          Mouse Genome 430 2.0 Array 1448621_PM_a_at - -
          Mouse Genome 430 2.0 Array 1448621_a_at - -
          Mouse Genome 430 2.0 Array 1447874_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20597_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20597_at - -
          Affymetrix Mouse Clariom S Array PSR0700012979.mm.1 TC0700001554.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012979.mm.1 NM_011421.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012979.mm.1 NM_011421.2 Refseq
          Mouse Expression Set 430 1448621_a_at - -
          Mouse Expression Set 430 1448621_PM_a_at - -
          Mouse Expression Set 430 1447874_x_at - -
          Mouse Expression Set 430 1447874_PM_x_at - -
          Murine Genome U74v2 Set 100099_at - -
          Murine Genome U74v2 Set 161789_PM_r_at - -
          Murine Genome U74v2 Set 100099_PM_at - -
          Murine Genome U74v2 Set 161789_r_at - -
          Mouse Genome 430A 2.0 Array 1448621_a_at - -
          Mouse Genome 430A 2.0 Array 1448621_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1448621_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1448621_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1447874_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1447874_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 869360 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5550717 - -
          Illumina mouse-6 v1.0 expression beadchip 1410309 - -
          Illumina mouse-6 v1.0 expression beadchip 5550717 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005550717 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001410309 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2728521 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1245356 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2728521 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245356 - -
          mouse-6 v1.1 (Illumina) scl0003.1_30-S - -
          mouse-6 v1.1 (Illumina) scl32240.5.1_5-S - -
          Illumina Sentrix mouse-6.v1 bead array 1410309 - -
          Illumina Sentrix mouse-6.v1 bead array 5550717 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0003.1_30-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32240.5.1_5-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5550717 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1410309 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1245356 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2868987 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2728521 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2728521 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2868987 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1245356 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl32240.5.1_5-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0003.1_30-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline